Skip to Content
Merck
All Photos(4)

Key Documents

I1754

Sigma-Aldrich

Isobutyric acid

99%

Synonym(s):

2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2H
CAS Number:
Molecular Weight:
88.11
Beilstein:
635770
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.04 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

824 °F

expl. lim.

10 %

refractive index

n20/D 1.393 (lit.)

bp

153-154 °C (lit.)

mp

−47 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(O)=O

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isobutyric acid is a carboxylic acid used as a building block in the production of active pharmaceutical ingredients (APIs).

Application

Isobutyric acid can be used:
  • to synthesize butyl isobutyrate via esterification with n-butanol using lipases
  • as a precursor for Methacrylic acid (MAA) through catalytic oxidative dehydrogenation. It can be produced through fermentative pathways

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Livia H Morais et al.
Current biology : CB, 30(19), 3761-3774 (2020-08-22)
Birth by Caesarean (C)-section impacts early gut microbiota colonization and is associated with an increased risk of developing immune and metabolic disorders. Moreover, alterations of the microbiome have been shown to affect neurodevelopmental trajectories. However, the long-term effects of C-section
E M Faed et al.
Clinical and experimental pharmacology & physiology, 5(2), 195-198 (1978-03-01)
1. Two main conjugates of CPIB (2-[chlorophenoxy]-2-methylpropionic acid) are present in the urine of subjects taking clofibrate. The metabolites can be separated by thin-layer chromatography (TLC). 2. Both conjugates are hydrolysed by dilute alkali, but only one is hydrolysed by
A Vyalikh et al.
Physical chemistry chemical physics : PCCP, 9(18), 2249-2257 (2007-05-10)
Solid state deuterium NMR has been used to study the molecular motion of d(6)-isobutyric acid (d(6)-iBA) in the pure (unconfined) state and confined in the cylindrical pores of two periodic mesoporous silica materials (MCM-41, pore size 3.3 nm and SBA-15
Ritu Lal et al.
The Journal of pharmacology and experimental therapeutics, 330(3), 911-921 (2009-06-09)
Baclofen is a racemic GABA(B) receptor agonist that has a number of significant pharmacokinetic limitations, including a narrow window of absorption in the upper small intestine and rapid clearance from the blood. Arbaclofen placarbil is a novel transported prodrug of
Julia Biermann et al.
Molecular vision, 17, 395-403 (2011-02-12)
Histone deacetylase inhibitors (HDACi) have neuroprotective effects under various neurodegenerative conditions, e.g., after optic nerve crush (ONC). HDACi-mediated protection of central neurons by increased histone acetylation has not previously been demonstrated in rat retinal ganglion cells (RGCs), although epigenetic changes

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service