Skip to Content
Merck
All Photos(1)

Key Documents

46417

Supelco

Flumethrin

PESTANAL®, analytical standard

Synonym(s):

Cyano(4-fluoro-3-phenoxyphenyl)methyl 3-[2-chloro-2-(4-chlorophenyl)ethenyl]-2,2-dimethylcyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22Cl2FNO3
CAS Number:
Molecular Weight:
510.38
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(\C=C(/Cl)c2ccc(Cl)cc2)C1C(=O)OC(C#N)c3ccc(F)c(Oc4ccccc4)c3

InChI

1S/C28H22Cl2FNO3/c1-28(2)21(15-22(30)17-8-11-19(29)12-9-17)26(28)27(33)35-25(16-32)18-10-13-23(31)24(14-18)34-20-6-4-3-5-7-20/h3-15,21,25-26H,1-2H3/b22-15-

InChI key

YXWCBRDRVXHABN-JCMHNJIXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flumethrin is a synthetic pyrethroid insecticide.

Application

Flumethrin may be used as a reference standard in the determination of flumethrin blood and milk samples of lactating dairy cows using high performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Sommer et al.
Bulletin of entomological research, 91(2), 131-137 (2001-03-22)
Dung from calves treated with synthetic pyrethroids negatively influenced, in varying degrees, survival, reproduction and size of the common dung fly Neomyia cornicina (Fabricius). This was documented in assays where the coprophagous larvae and adults of N. cornicina were exposed
Josephus J Fourie et al.
Parasites & vectors, 5, 151-151 (2012-07-31)
The objective of the study was to determine the sustained effectiveness of 10% imidacloprid (w/w) and 4.5% flumethrin (w/w) incorporated in a slow-release matrix collar in preventing Dipylidium caninum infection in cats following repeated laboratory-infestations with fleas infected with metacestodes.
S Shimizu et al.
Veterinary parasitology, 92(2), 129-138 (2000-08-18)
The effectiveness of the pour-on formulation of flumethrin was tested on grazing cattle. Flumethrin was applied once a month from April to October from 1990 to 1995 to cattle grazing in the Aso area of Kumamoto Prefecture in Japan. Both
I Floris et al.
Journal of economic entomology, 94(4), 806-810 (2001-09-20)
An apiary trial was conducted in 1997 in Sardinia, Italy, to verify the effectiveness of fluvalinate in polyvinyl chloride strips and flumethrin in polyethylene strips against Varroa jacobsoni Oudemans. Two indices to evaluate the efficacy of the treatments were adopted:
HPLC determination of flumethrin, deltamethrin, cypermethrin, and cyhalothrin residues in the milk and blood of lactating dairy cows.
Bissacot Z D and Vassilieff I.
Journal of Analytical Toxicology, 21(5), 397-402 (1997)

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service