Skip to Content
Merck
All Photos(1)

Documents

45806

Supelco

α-Cypermethrin

PESTANAL®, analytical standard

Synonym(s):

α-metherin, Alphamethrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
Beilstein:
8444118
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Manoch Naksata et al.
International journal of environmental research and public health, 17(9) (2020-05-14)
Wearing appropriate personal protective equipment during the application of pesticides is one method of reducing dermal exposure to pesticides. Thus, the aim of this research is to develop personal protective clothing (PPC) coated with gum rosin and investigate the efficiency
George P Lahm et al.
Bioorganic & medicinal chemistry letters, 15(22), 4898-4906 (2005-09-17)
A novel class of anthranilic diamides has been discovered with exceptional insecticidal activity on a range of Lepidoptera. These compounds have been found to exhibit their action by release of intracellular Ca2+ stores mediated by the ryanodine receptor. The discovery
María Carla Cecere et al.
Journal of medical entomology, 50(2), 394-403 (2013-04-02)
The effectiveness of two doses of suspension concentrate (SC) pyrethroid insecticides in suppressing peridomestic populations of Triatoma infestans (Klug) was evaluated in 28 rural communities located in Santiago del Estero province, northwestern Argentina, including 388 houses and 1,516 identified sites.
Valentina Sechi et al.
Environmental pollution (Barking, Essex : 1987), 184, 586-596 (2013-11-10)
A soil multi-species, SMS, experimental test system consisting of the natural microbial community, five collembolan species and a predatory mite along with either Enchytraeus crypticus or the earthworm Eisenia fetida were exposed to α-cypermethrin. A comparison of the performance of
M L Groner et al.
Journal of fish diseases, 36(3), 195-208 (2012-11-06)
Sea lice, Lepeophtheirus salmonis, are ectoparasites of farmed and wild salmonids. Infestations can result in significant morbidity and mortality of hosts in addition to being costly to control. Integrated pest management programmes have been developed to manage infestations, and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service