Skip to Content
Merck
All Photos(3)

Documents

I8898

Sigma-Aldrich

Ivermectin

Synonym(s):

22,23-Dihydroavermectin B1, MK-933

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

solubility

MEK: 50 mg/mL

antibiotic activity spectrum

parasites

application(s)

agriculture
environmental

Mode of action

cell membrane | interferes

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

[H][C@@]1(O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C6/CO[C@]7([H])[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]67O)O2)[C@@H](C)CC

InChI

1S/C48H74O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,19,25-26,28,30-31,33-45,49-50,52H,11,16-18,20-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1

InChI key

AZSNMRSAGSSBNP-XPNPUAGNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Ivermectin is a macro-cyclic lactone, derived from avermectin. It functions as an antiparasitic agent. Ivermectin is used against gastrointestinal roundworms, lungworms, mites and lice. It is used to treat onchocerciasis, strongyloidiasis, ascariasis and filariases.

Application

Ivermectin has been used:
  • in Caenorhabditis elegans fecundity assay
  • in larval development test
  • to measure in vivo topical acaricide activity of compounds in rats
  • to assess its acute and chronic effects to zebrafish
  • in percutaneous drug injection method to treat cystic echinococcosis
  • in liquid chromatography with fluorescence detection method

Biochem/physiol Actions

Positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptor; also modulates glutamate-GABA-activated chloride channels.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Primarily ivermectin B1a.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Martin et al.
International journal for parasitology, 28(6), 849-862 (1998-07-23)
Two of three major types of anthelminitic, the avermectins and the nicotinic agonists, exert their therapeutic effect by an action on ligand-gated membrane ion-channels of nematodes. The avermectins, such as ivermectin, open glutamategated chloride channels which have so far been
Ivermectin in human medicine
Ottesen EA and Campbell W
The Journal of Antimicrobial Chemotherapy, 34(2), 195-203 (1994)
Ivermectin used in percutaneous drug injection method for the treatment of liver hydatid disease in sheep
Hokelek M, et al.
Gastroenterology, 122(4), 957-962 (2002)
Evaluation of the toxicity of 2-aminoimidazole antibiofilm agents using both cellular and model organism systems
Stowe SD, et al.
Drug and Chemical Toxicology, 35(3), 310-315 (2012)
R M Krause et al.
Molecular pharmacology, 53(2), 283-294 (1998-03-14)
We report that preapplication of ivermectin, in the micromolar range, strongly enhances the subsequent acetylcholine-evoked current of the neuronal chick or human alpha7 nicotinic acetylcholine receptors reconstituted in Xenopus laevis oocytes and K-28 cells. This potentiation does not result from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service