Skip to Content
Merck
All Photos(1)

Key Documents

45686

Supelco

Thidiazuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N4OS
CAS Number:
Molecular Weight:
220.25
Beilstein:
1078092
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(Nc1ccccc1)Nc2cnns2

InChI

1S/C9H8N4OS/c14-9(12-8-6-10-13-15-8)11-7-4-2-1-3-5-7/h1-6H,(H2,11,12,14)

InChI key

HFCYZXMHUIHAQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thidiazuron is a systemic herbicide, widely employed as an effective plant growth regulator and pre-harvest defoliant for crops such as cotton.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thidiazuron may be used as an analytical reference standard for the quantification of the analyte in fruits, vegetables and environmental samples using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Selective determination of thidiazuron herbicide in fruit and vegetable samples using molecularly imprinted polymer fiber solid phase microextraction with ion mobility spectrometry detection (MIPF-SPME-IMS)
Mirzajani.R, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 130, 93-101 (2017)
Application of dispersive liquid-liquid microextraction for the determination of phenylurea herbicides in water samples by HPLC-diode array detection
Saraji M and Tansazan N
Journal of Separation Science, 32(23-24), 4186-4192 (2009)
Lucas Cutri et al.
Mechanisms of development, 130(1), 61-69 (2012-06-05)
Tendrils can be found in different plant species. In legumes such as pea, tendrils are modified leaves produced by the vegetative meristem but in the grape vine, a same meristem is used to either form a tendril or an inflorescence.
M Cano-Castillo et al.
Cryo letters, 33(3), 182-190 (2012-07-25)
We cryopreserved in vitro shoot tips of saltcedar (Tamarix boveana Bunge) using the vitrification technique. The success of the cryopreservation protocol was strongly affected by preculture, loading duration, dehydration duration in plant vitrification solution 2 (PVS2), and medium composition during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service