Skip to Content
Merck
All Photos(1)

Key Documents

D5912

Sigma-Aldrich

6-(γ,γ-Dimethylallylamino)purine

BioReagent, suitable for plant cell culture, ≥90%

Synonym(s):

N6 dimethylallyladenine, 2iP, N6-(2-Isopentenyl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

Assay

≥90%

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(C)=C\CNc1ncnc2[nH]cnc12

InChI

1S/C10H13N5/c1-7(2)3-4-11-9-8-10(13-5-12-8)15-6-14-9/h3,5-6H,4H2,1-2H3,(H2,11,12,13,14,15)

InChI key

HYVABZIGRDEKCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-(γ,γ-Dimethylallylamino)purine: has been used as a standard in ultra-high performance liquid chromatography to compare the chromatogram of transgenic and wild type (WT) crude hormone extracts to quantify cytokinin levels, may be used as a component of the Murashige and Skoog (MS) medium for callus culture and in vitro shoot induction, may be used as a supplement in MS medium to make simplified shoot induction medium (SIM) to treat explants for shoot induction

Biochem/physiol Actions

6-(γ,γ-Dimethylallylamino)purine (2iP) is a bacteria-derived riboside cytokinin used to grow plant tissues such as tobacco and soybean callus. 2iP is a precursor of the cytokinin Zeatin. 2iP can also be used in Schenk and Hildebrandt medium to support in vitro propagation of micro shoot cultures from shoot tips of Genista plants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzislava Ckurshumova et al.
The New phytologist, 204(3), 556-566 (2014-10-03)
In vitro regeneration of complete organisms from diverse cell types is a spectacular property of plant cells. Despite the great importance of plant regeneration for plant breeding and biotechnology, its molecular basis is still largely unclear and many important crop
The development of an efficient cultivar-independent plant regeneration system from callus derived from both apical and non-apical root segments of garlic (Allium sativum L.)
Zheng SJ, et al
In Vitro Cellular & Developmental Biology. Plant, 39(3), 288-292 (2003)
E Strzelczyk et al.
Acta microbiologica Polonica, 34(2), 177-185 (1985-01-01)
Studies on the effect of post culture liquids of actinomycetes on cytokinin-like substances production by mycorrhizal fungi have revealed that actinomycete metabolites inhibited or stimulated the synthesis of these compounds. The results of chromatographic analyses suggest, that substances stimulating the
Maria Luczkiewicz et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 60(7-8), 557-566 (2005-09-17)
A two-stage method for in vitro propagation of six Genista species from shoot tips was developed. Multiple microshoot cultures were obtained by growing the shoot tip explants on Schenk and Hildebrandt medium supplemented with 9.84 microM 6-(gamma,gamma-dimethylallylamino)-purine and 0.99 microM
G A Miura et al.
Plant physiology, 44(3), 372-376 (1969-03-01)
A 6-(gamma,gamma-dimethylallylamino) purine-like compound was found in the culture medium of Rhizopogon roseolus, which had been shown earlier to synthesize zeatin. The role of 6-(gamma,gamma-dimethylallylamino) purine as a precursor of zeatin was studied. Rhizopogon was furnished with 6-(gamma,gamma-dimethylallylamino) purine-8-(14)C. Cochromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service