Skip to Content
Merck
All Photos(2)

Key Documents

45414

Supelco

Cyromazin

PESTANAL®, analytical standard

Synonym(s):

N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N6
CAS Number:
Molecular Weight:
166.18
Beilstein:
882879
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

223-227 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc(N)nc(NC2CC2)n1

InChI

1S/C6H10N6/c7-4-10-5(8)12-6(11-4)9-3-1-2-3/h3H,1-2H2,(H5,7,8,9,10,11,12)

InChI key

LVQDKIWDGQRHTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D B Taylor et al.
Journal of economic entomology, 105(2), 726-731 (2012-05-23)
Hay mixed with manure and urine residues at sites where hay has been provided as supplemental winter feed for cattle provide an excellent substrate for the development of immature stable flies, Stomoxys calcitrans (L.). Such sites are primary sources of
Kang Xia et al.
Journal of agricultural and food chemistry, 58(10), 5945-5949 (2010-02-17)
Cyromazine is used as an additive in poultry feed to inhibit the development of fly larvae in chicken manure. U.S. Environmental Protection Agency (EPA) method AG-555, modified from method AG-376, has been the standard method for cyromazine analysis in poultry
G W Levot
Australian veterinary journal, 91(1-2), 61-64 (2013-01-30)
To determine the significance of cyromazine resistance in blowfly larvae from a region where resistance has been previously confirmed and to measure susceptibility of the cyromazine-resistant blowflies to ivermectin and spinosad. Blowfly larvae from five properties located within 5 km
Xianhua Wang et al.
Analytical and bioanalytical chemistry, 404(5), 1555-1564 (2012-07-24)
A magnetic molecularly imprinted polymer (M-MIP) for cyromazine and melamine was prepared by simple suspension polymerization using a pseudo template, 2-(4,6-diamino-1,3,5-triazin-2-ylamino)ethanethiol disulfide. The M-MIP was characterized by scanning electron microscopy, Fourier-transform infrared spectroscopy, and vibrating sample magnetometry. Molecular recognition properties
Hanwen Sun et al.
Electrophoresis, 31(13), 2236-2241 (2010-07-02)
A novel and simple method was, for the first time, developed for effective separation and sensitive detection of cyromazine (CM) and its metabolite melamine (MM) by CZE with DAD (CZE-DAD). Among different solvents, ACN containing 20% NH(4)OH was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service