Skip to Content
Merck
All Photos(1)

Key Documents

45511

Supelco

Fluridon

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14F3NO
CAS Number:
Molecular Weight:
329.32
Beilstein:
1547990
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN1C=C(c2ccccc2)C(=O)C(=C1)c3cccc(c3)C(F)(F)F

InChI

1S/C19H14F3NO/c1-23-11-16(13-6-3-2-4-7-13)18(24)17(12-23)14-8-5-9-15(10-14)19(20,21)22/h2-12H,1H3

InChI key

YWBVHLJPRPCRSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Rasmussen et al.
Plant signaling & behavior, 7(6), 694-697 (2012-05-15)
Roots that form from non-root tissues (adventitious roots) are crucial for cutting propagation in the forestry and horticulture industries. Strigolactone has been demonstrated to be an important regulator of these roots in both Arabidopsis and pea using strigolactone deficient mutants
Woong Han et al.
BMB reports, 43(12), 813-817 (2010-12-30)
Plants undergo cell division throughout their life in order to maintain their growth. It is well known that root and shoot tip of plants possess meristems, which contain quiescent cells. Fluridone (1-methyl-3-phenyl-5-(3-trifluoromethyl (phenyl))-4-(1H)-pyridinone) is an established inhibitor of both ABA
Xue-Yi Qiao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 32(18), 1848-1850 (2007-12-07)
To study the effect of fluridone concentration, stimulating period, temperature and salt on the seed germination of three species of Cistanche. The seeds were cultured in Petri dish, and the germination percentage was counted. The highest germination percentage was observed
A Ben Hassine et al.
Annals of botany, 104(5), 925-936 (2009-08-12)
Different populations of the Mediterranean xerohalophyte species Atriplex halimus exhibit different levels of resistance to salt and osmotic stress depending on the nature of the osmocompatible solute they accumulate. There is, however, no conclusive description of the involvement of abscisic
Lina Sun et al.
Plant cell reports, 31(1), 179-185 (2011-09-29)
Exposure to ozone induced a rapid increase in the levels of the sesquiterpene phytohormone abscisic acid (ABA) and the isoflavone puerarin in suspension cell cultures of Pueraria thomsnii Benth. The observed increases in ABA and puerarin were dependent on the

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the IUPAC name for Product 45511, Fluridon?

    1 answer
    1. The IUPC name for Product 45511, Fluridon is 4(1H)-Pyridinone, 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl or 1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]-4(1H)-pyridinone.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service