Skip to Content
Merck
All Photos(4)

Key Documents

C0143

Sigma-Aldrich

Ethephon

≥96% (titration)

Synonym(s):

2-Chloroethylphosphonic acid, Ethephon

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
144.49
Beilstein:
1751208
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

Assay

≥96% (titration)

form

solid

mp

70-72 °C (lit.)

application(s)

agriculture

storage temp.

2-8°C

SMILES string

OP(O)(=O)CCCl

InChI

1S/C2H6ClO3P/c3-1-2-7(4,5)6/h1-2H2,(H2,4,5,6)

InChI key

UDPGUMQDCGORJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethephon (2-Chloroethylphosphonic acid), a widely used plant growth regulator that induces scenescence and fruiting may be used to study its properties as a plant growth regulator, phosphorylating agent and as a butyrylcholinesterase inhibitor. Ethephon may be used as a reference compound in procedures designed to detect ethephon in biological materials.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paiboon Tunsagool et al.
Plant cell reports, 38(5), 559-575 (2019-02-05)
Bacillus subtilis CLP extract activates defense gene expression and increases the unique protein production involving in pathways of ISR, SAR, ubiquitin-proteasome system, and glycolysis for stress responses in flavedo tissues. Cyclic lipopeptides (CLPs) of Bacillus subtilis ABS-S14 had ability to
Hsien-Jung Chen et al.
Journal of plant physiology, 169(1), 86-97 (2011-09-07)
In this report a full-length cDNA, SPCAT1, was isolated from ethephon-treated mature L3 leaves of sweet potato. SPCAT1 contained 1479 nucleotides (492 amino acids) in its open reading frame, and exhibited high amino acid sequence identities (ca. 71.2-80.9%) with several
Shigemi Seo et al.
Plant physiology, 155(1), 502-514 (2010-11-16)
Rice (Oryza sativa) plants carrying the Pi-i resistance gene to blast fungus Magnaporthe oryzae restrict invaded fungus in infected tissue via hypersensitive reaction or response (HR), which is accompanied by rapid ethylene production and formation of small HR lesions. Ethylene
J E Haux et al.
Chemical research in toxicology, 13(7), 646-651 (2000-07-18)
Ethephon [(2-chloroethyl)phosphonic acid] has two seemingly unrelated types of biological activity. It is a major agrochemical absorbed by crops, slowly releasing ethylene as a plant growth regulator. Ethephon also inhibits the activity of plasma butyrylcholinesterase (BuChE) in humans, dogs, rats
Enriqueta Moyano-Cañete et al.
Plant & cell physiology, 54(2), 218-236 (2012-12-13)
Numerous GAST-like genes have been reported in higher plants, but only one GAST-like gene (FaGAST1) has been described in strawberry so far. Herein, we have identified a novel strawberry FaGAST gene (FaGAST2) whose expression showed an increase throughout fruit receptacle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service