Skip to Content
Merck
All Photos(1)

Documents

38960

Sigma-Aldrich

Dimethylamine hydrochloride

purum, ≥98.0% (AT)

Synonym(s):

N,N-Dimethylamine hydrochloride, N-Methylmethanamine hydrochloride, Dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH · HCl
CAS Number:
Molecular Weight:
81.54
Beilstein:
3589311
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (AT)

impurities

≤2% water

mp

170-173 °C (lit.)

solubility

H2O: 0.5 g/10 mL

SMILES string

Cl[H].CNC

InChI

1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H

InChI key

IQDGSYLLQPDQDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Zhang et al.
Clinical biochemistry, 45(13-14), 1064-1069 (2012-05-23)
To develop a noninvasive and accessible diagnostic method for pancreatic cancer (PC). We presented a metabolomic method, pattern recognition techniques applied to (1)H nuclear magnetic resonance ((1)H NMR) spectra, to investigate the plasma metabolites obtained from 19 patients with PC
Peter I Nagy
Physical chemistry chemical physics : PCCP, 14(2), 849-857 (2011-11-24)
Monte Carlo simulations have been performed for characterizing the 0.22 and 1 molar aqueous dimethylammonium chloride solutions at p = 1 atm and T = 310 K. On the basis of potential of mean force curves for the two systems
Patrick L Feng et al.
Dalton transactions (Cambridge, England : 2003), 41(29), 8869-8877 (2012-06-19)
One of the ongoing goals in the field of porous materials is the design and synthesis of materials that possess chemical structures amenable for use in sensing applications. We describe the preparation, luminescence characteristics, and environmental sensing properties of variants
Guozhen Liu et al.
Analytical chemistry, 84(9), 3921-3928 (2012-03-28)
A glassy carbon substrate was covalently modified with a mixed layer of 4-aminophenyl and phenyl via in situ electrografting of their aryldiazonium salts in acidic solutions. Single-walled carbon nanotubes (SWNTs) were covalently and vertically anchored on the electrode surface via
Lokesh Padhye et al.
Environmental science & technology, 45(10), 4353-4359 (2011-04-21)
Interactions of ozone with organic precursors during water treatment may generate carcinogenic N-nitrosodimethylamine (NDMA) byproduct. This study investigates the reaction mechanisms responsible for NDMA formation from ozonation of the commonly used poly(diallyldimethylammonium chloride) (polyDADMAC) coagulant. Upon ozonation, polyDADMAC yields the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service