Skip to Content
Merck
All Photos(1)

Key Documents

36576

Supelco

Sulfur

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

product line

PESTANAL®

autoignition temp.

450 °F

shelf life

limited shelf life, expiry date on the label

resistivity

2E23 μΩ-cm, 20°C

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur can play the role as a cathode material and can also find applications in electric and hybrid electric vehicles. It can also form a composite with graphite, which can serve as an improved sulfur cathode materials for lithium-sulfur(Li-S) batteries with higher capacity and cycling stability.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-Quan Liu et al.
Science advances, 4(10), eaat5383-eaat5383 (2018-10-13)
High-energy lithium metal batteries (LMBs) are expected to play important roles in the next-generation energy storage systems. However, the uncontrolled Li dendrite growth in liquid electrolytes still impedes LMBs from authentic commercialization. Upgrading the traditional electrolyte system from liquid to
Graphene-wrapped sulfur particles as a rechargeable lithium?sulfur battery cathode material with high capacity and cycling stability
Wang H, et al.
Nano Letters, 11, 2644-2647 (2011)
Porous hollow carbon@ sulfur composites for high?power lithium?sulfur batteries
Jayaprakash.N, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 123, 6026-6030 (2011)
Farhad Forouhar et al.
Nature chemical biology, 9(5), 333-338 (2013-04-02)
How living organisms create carbon-sulfur bonds during the biosynthesis of critical sulfur-containing compounds is still poorly understood. The methylthiotransferases MiaB and RimO catalyze sulfur insertion into tRNAs and ribosomal protein S12, respectively. Both belong to a subgroup of radical-S-adenosylmethionine (radical-SAM)
Chemoselective peptide ligation-desulfurization at aspartate.
Robert E Thompson et al.
Angewandte Chemie (International ed. in English), 52(37), 9723-9727 (2013-07-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service