Skip to Content
Merck
All Photos(1)

Key Documents

451924

Sigma-Aldrich

Sulfur-32S

99.9 atom % 32S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
32S
CAS Number:
Molecular Weight:
31.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99.9 atom % 32S

mp

119-122 °C (lit.)

application(s)

environmental

mass shift

depleted

SMILES string

S

InChI

1S/S/i1+0

InChI key

NINIDFKCEFEMDL-IGMARMGPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Polhemus et al.
Cardiovascular therapeutics, 33(4), 216-226 (2015-05-02)
Recent studies demonstrate robust molecular cross talk and signaling between hydrogen sulfide (H2 S) and nitric oxide (NO). Heart failure (HF) patients are deficient in both H2 S and NO, two molecules that are critical for cardiovascular homeostasis. A phase
Haitao Shi et al.
Journal of integrative plant biology, 57(7), 628-640 (2014-10-21)
Hydrogen sulfide (H2S) is an important gaseous molecule in various plant developmental processes and plant stress responses. In this study, the transgenic Arabidopsis thaliana plants with modulated expressions of two cysteine desulfhydrases, and exogenous H2S donor (sodium hydrosulfide, NaHS) and
V R G Mercadante et al.
Journal of animal science, 93(1), 414-424 (2015-01-09)
In Exp. 1, individual performance and daily DMI was measured on 70 crossbred weaned calves during a 70-d period using a GrowSafe system (GrowSafe Systems Ltd., Airdrie, AB, Canada) at the University of Florida North Florida Research and Education Center
Beata Olas et al.
Chemico-biological interactions, 220, 20-24 (2014-06-15)
Hydrogen sulfide (H2S) is a well known toxic gas at high levels. However, at physiological levels, H2S may play a role in the pathogenesis of various cardiovascular diseases. The objective was to study the effects of exogenous H2S on the
Fadi N Salloum et al.
Experimental biology and medicine (Maywood, N.J.), 240(5), 669-681 (2014-11-02)
Ingestion of high dietary nitrate in the form of beetroot juice (BRJ) has been shown to exert antihypertensive effects in humans through increasing cyclic guanosine monophosphate (cGMP) levels. Since enhanced cGMP protects against myocardial ischemia-reperfusion (I/R) injury through upregulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service