Skip to Content
Merck
All Photos(1)

Key Documents

34179

Supelco

Imazapic

PESTANAL®, analytical standard

Synonym(s):

2-(4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methylnicotinic acid, 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-methyl-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N3O3
CAS Number:
Molecular Weight:
275.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C1(C)N=C(NC1=O)c2ncc(C)cc2C(O)=O

InChI

1S/C14H17N3O3/c1-7(2)14(4)13(20)16-11(17-14)10-9(12(18)19)5-8(3)6-15-10/h5-7H,1-4H3,(H,18,19)(H,16,17,20)

InChI key

PVSGXWMWNRGTKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazapic is an imidazolinone herbicide which is effective against weedy rice constraints.

Application

Imazapic may be used as a reference standard for the determination of imazapic in rice samples by high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wangcang Su et al.
PloS one, 14(7), e0219462-e0219462 (2019-07-10)
Imazapic is widely used in peanut production, and its residues can cause damage to succeeding crops planted in the following year. The planting area of peanut is large in Henan province. Inceptisol is the main soil type in Henan Province
S Stone et al.
Environmental pollution (Barking, Essex : 1987), 250, 97-106 (2019-04-16)
Microalgae are commonly used in ecotoxicity testing due to their ease of culturing and rapid cell division rates. These tests generally utilise a single species of algae; however, microalgae occur in the environment as complex communities of multiple species. To
Determination of residues of imazapic and imazaphic in rice (Oryza sativa) by HPLC.
Silva T.
Ingenieria, 15(1-2) (2005)
Shahar Pincovici et al.
Plant science : an international journal of experimental plant biology, 271, 100-107 (2018-04-14)
Sunflower broomrape (Orobanche cumana) is a root holoparasitic plant causing major damage to sunflower (Helianthus annuus L.). Parasite infection initiates source-sink relations between the parasite (sink) and the host (source), allocating carbohydrates, water and nutrients to the parasite. The primary
Seth M Munson et al.
Ecological applications : a publication of the Ecological Society of America, 30(7), e02151-e02151 (2020-04-29)
Restoration and rehabilitation are globally implemented to improve ecosystem condition but often without tracking treatment expenditures relative to ecological outcomes. We evaluated the cost-effectiveness of widely conducted woody plant and herbaceous invasive plant removals and seeding treatments in drylands of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service