Skip to Content
Merck
All Photos(1)

Key Documents

45817

Supelco

Haloxyfop

PESTANAL®, analytical standard

Synonym(s):

2-[4-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]propanoic acid, Haloxyfop acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Beilstein:
1507817
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1)C(O)=O

InChI

1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)

InChI key

GOCUAJYOYBLQRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Banaś et al.
Biochemical Society transactions, 28(6), 777-779 (2001-02-15)
Seedlings of wheat were grown for 24 h in control nutrient solution and in solutions containing haloxyfop, alloxydim, diquat or paraquat, and thereafter the roots were used for microsomal preparations. Phosphatidylcholine or diacylglycerol with various 1-(14)C-labelled fatty acids (oleic, linoleic
P Gornicki et al.
Plant molecular biology, 22(3), 547-552 (1993-06-01)
The acetyl-CoA carboxylase present in both wheat germ and total wheat leaf protein contains ca. 220 kDa subunits. It is the major biotin-dependent carboxylase present in wheat chloroplasts. Active acetyl-CoA carboxylase purified from wheat germ is a homodimer with an
Hailong Zhang et al.
Science (New York, N.Y.), 299(5615), 2064-2067 (2003-03-29)
Acetyl-coenzyme A carboxylases (ACCs) are required for the biosynthesis and oxidation of long-chain fatty acids. They are targets for therapeutics against obesity and diabetes, and several herbicides function by inhibiting their carboxyltransferase (CT) domain. We determined the crystal structure of
M J Bartels et al.
Drug metabolism and disposition: the biological fate of chemicals, 17(3), 286-291 (1989-05-01)
The 2-aryloxypropionate haloxyfop is currently being evaluated for use as a herbicide. This compound is structurally similar to a group of 2-arylpropionates that have been shown previously to undergo stereochemical inversion in a variety of mammalian species. To support the
W T Stott et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 28(1), 71-79 (1995-11-01)
The potential of haloxyfop [2-(4-((3-chloro-5-(trifluoromethyl)-2- pyridinyl)oxy)phenoxy)propanoic acid; HAL] to induce the proliferation of hepatocellular peroxisomes (PP) was examined in rats, mice, dogs, and monkeys. Chemically induced PP is associated with the development of liver tumors in rodents via an apparent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service