Skip to Content
Merck
All Photos(1)

Key Documents

33993

Supelco

Doramectin

VETRANAL®, analytical standard

Synonym(s):

25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H74O14
CAS Number:
Molecular Weight:
899.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12OC/C3=C\C=C\[C@H](C)[C@H](O[C@@H]4C[C@H](OC)[C@@H](O[C@@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)\C(C)=C\CC6C[C@@H](C[C@]7(O6)O[C@@H]([C@@H](C)C=C7)C8CCCCC8)OC(=O)[C@H](C=C(C)[C@H]1O)[C@@]23O

InChI

1S/C50H74O14/c1-27-13-12-16-34-26-57-47-42(51)30(4)21-37(50(34,47)54)48(53)60-36-22-35(63-49(25-36)20-19-29(3)45(64-49)33-14-10-9-11-15-33)18-17-28(2)44(27)61-41-24-39(56-8)46(32(6)59-41)62-40-23-38(55-7)43(52)31(5)58-40/h12-13,16-17,19-21,27,29,31-33,35-47,51-52,54H,9-11,14-15,18,22-26H2,1-8H3/b13-12+,28-17+,34-16+/t27-,29-,31-,32-,35?,36-,37-,38-,39-,40+,41+,42+,43-,44-,45-,46-,47+,49+,50+/m0/s1

InChI key

QLFZZSKTJWDQOS-TTXKVKCHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gokbulut et al.
Laboratory animals, 44(2), 138-142 (2009-12-05)
This study evaluated the comparative plasma dispositions of ivermectin (IVM), doramectin (DRM) and moxidectin (MXD) following subcutaneous administration in rabbits. Fifteen New Zealand white rabbits were allocated into three groups of five animals each. The animals in each group received
Efficacy of weekly oral doramectin treatment in canine demodicosis.
N Murayama et al.
The Veterinary record, 167(2), 63-64 (2010-07-14)
Jan Antonić et al.
Veterinary parasitology, 179(1-3), 159-166 (2011-04-07)
The aim of to the present study was to evaluate the effects of verapamil (VER) on plasma pharmacokinetics of ivermectin (IVM) and doramectin (DOR) in lactating Istrian Pramenka dairy sheep and to investigate the role of P-glycoprotein (P-gp) in transport
Hamza Avcioglu et al.
Tropical animal health and production, 43(6), 1097-1099 (2011-02-26)
This study was conducted to investigate the efficacy of ivermectin (IVM), doramectin (DRM), and moxidectin (MXD) against Toxocara vitulorum in calves. In the study, 20 calves naturally infected with T. vitulorum were divided into four groups: three different treatment groups
Ginger L Takle et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 41(3), 496-502 (2010-10-16)
One female and two male 4-mo-old, sibling rock hyraxes (Procavia capensis) presented with severe generalized dermatitis characterized by nonpruritic, nonalopecic, mildly encrusted, focally, ulcerated pustular nodules. The dorsum, limbs, and dorsal cranium were most severely affected. Skin scrapings, histopathology, and

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service