Skip to Content
Merck
All Photos(1)

Key Documents

75100

Sigma-Aldrich

Ethyl oleate

tested according to Ph. Eur.

Synonym(s):

Ethylis oleas, Oleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Agency

tested according to Ph. Eur.

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

216-218 °C/15 mmHg

mp

−32 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-

InChI key

LVGKNOAMLMIIKO-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl oleate can be used as a model for fats to study their degree of ozonization and decomposition enthalpy of ozonides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical and thermochemical aspects of the ozonolysis of ethyl oleate: Decomposition enthalpy of ethyl oleate ozonide.
Cataldo F.
Chemistry and Physics of Lipids, 175, 41-49 (2013)
Michael D Hardy et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8187-8192 (2015-06-24)
Cell membranes are dynamic structures found in all living organisms. There have been numerous constructs that model phospholipid membranes. However, unlike natural membranes, these biomimetic systems cannot sustain growth owing to an inability to replenish phospholipid-synthesizing catalysts. Here we report
Shujauddin Changi et al.
ChemSusChem, 5(9), 1743-1757 (2012-08-29)
We examined the behavior of phenylalanine in high-temperature water (HTW) at 220, 250, 280, and 350 °C. Under these conditions, the major product is phenylethylamine. The minor products include styrene and phenylethanol (1-phenylethanol and 2-phenylethanol), which appear at higher temperatures and
Y Ingrid Goh et al.
Alcohol (Fayetteville, N.Y.), 44(7-8), 629-634 (2010-06-30)
Meconium fatty acid ethyl esters (FAEEs) are sensitive and specific biomarkers for prenatal alcohol exposure (PAE) in pregnancy. We recently reported a 2.5% rate of FAEE positive meconium in a general population sample of infants born in the region of
Aihua Yu et al.
Journal of pharmaceutical sciences, 100(3), 933-941 (2010-09-24)
The main objective of the study was to investigate the efficacy of microemulsion (ME) to facilitate bioavailability of puerarin (PUE) after oral and nasal administration. The pseudo-ternary phase diagrams were constructed to screen the ME components and optimize the ME

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service