Skip to Content
Merck
All Photos(1)

Documents

32955

Supelco

4,4′-Diaminodiphenyl sulfide

analytical standard

Synonym(s):

4,4′-Thiodianiline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(C6H4NH2)2
CAS Number:
Molecular Weight:
216.30
Beilstein:
1875513
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

105-107 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(Sc2ccc(N)cc2)cc1

InChI

1S/C12H12N2S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

ICNFHJVPAJKPHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4,4'-Thiodianiline.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 147-154 (1982-04-01)
L Pomorski et al.
Neoplasma, 49(4), 247-250 (2002-10-17)
Male Wistar rats were divided into two groups. Rats of group 1 were fed basal powdered diet containing 610 ppm 2,4- diaminoanisole sulfate (DAAS), 46 ppm 4,4'-thiodianiline (TDA) and 200 ppm N,N'-diethylthiourea (DETU) for 52 weeks (DTD treatment). Rats of
4,4'-Thiodianiline.
Report on carcinogens : carcinogen profiles, 12, 404-405 (2011-08-25)
4,4'-Thiodianiline.
Report on carcinogens : carcinogen profiles, 11, III248-III249 (2004-01-01)
Francesca Mattioli et al.
Toxicology and applied pharmacology, 203(2), 99-105 (2005-02-16)
Four chemicals that are known to induce in rats thyroid follicular-cell adenomas and carcinomas were assayed for their ability to induce DNA damage and DNA repair synthesis in primary cultures of human thyroid cells. Significant dose-dependent increases in the frequency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service