Skip to Content
Merck
All Photos(1)

Key Documents

33658

Supelco

Picoxystrobin

PESTANAL®, analytical standard

Synonym(s):

Methyl (E)-α-methoxymethylene-2-(3-trifluoromethyl-2-pyridyloxymethyl)phenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16F3NO4
CAS Number:
Molecular Weight:
367.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CO\C=C(/C(=O)OC)c1ccccc1COc2cccc(n2)C(F)(F)F

InChI

1S/C18H16F3NO4/c1-24-11-14(17(23)25-2)13-7-4-3-6-12(13)10-26-16-9-5-8-15(22-16)18(19,20)21/h3-9,11H,10H2,1-2H3/b14-11-

InChI key

IBSNKSODLGJUMQ-KAMYIIQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Picoxystrobin is classified under the strobulins family of fungicides that exhibit broad range activity against plant pathogenspathogenic fungi.

Application

Picoxystrobin may be used as a reference standard for the determination of picoxystrobin in:
  • Honeybees using acetonitrile-based extraction followed by dispersive solid-phase extraction (d-SPE) cleanup and gas chromatography−tandem quadrupole mass spectrometry (GC−MS/MS).
  • Foodstuffs by liquid chromatography (LC) combined with MS/MS, atmospheric pressure chemical ionization (APCI) and electrospray ionization (ESI).
  • Paddy field water by direct injection using LC coupled with quadrupole-linear ion trap (QqLIT)-MS.
  • Maize by acetonitrile-based extraction with dispersive liquid−liquid microextraction (DLLME) followed by GC-MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinzhong Zhang et al.
Journal of the science of food and agriculture, 101(1), 194-204 (2020-07-06)
Picoxystrobin is a new osmotic and systemic broad-spectrum methoxyacrylate fungicide with a good control effect on tea anthracnose, so it has been proposed to spray picoxystrobin before the occurrence and onset of tea anthracnose during tea bud growth in order
Honglei Zhao et al.
Journal of the science of food and agriculture, 100(14), 5145-5151 (2020-06-11)
Pyraclostrobin and picoxystrobin are two representative pesticides of strobilurins used to treat cucumber downy mildew, which have raised issues of food safety and human health. A new formulation containing these two compounds is being prepared for marketing in China. The
Marianne Stenrød et al.
PloS one, 8(6), e66989-e66989 (2013-07-03)
There is widespread use of chemical amendments to meet the demands for increased productivity in agriculture. Potentially toxic compounds, single or in mixtures, are added to the soil medium on a regular basis, while the ecotoxicological risk assessment procedures mainly
Yabing Duan et al.
Pesticide biochemistry and physiology, 153, 95-105 (2019-02-13)
Target leaf spot caused by Corynespora cassiicola is an economically important foliar disease on cucumber. In recent years, this disease has caused a serious problem on greenhouse-cultivated cucumber in China. In this study, to explore the characteristics and possible causes
The ECHO technique-the more effective way of data evaluation in liquid chromatography-tandem mass spectrometry analysis
Alder L, et al.
Journal of Chromatography A, 1058(1-2), 67-79 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service