Skip to Content
Merck
All Photos(1)

Key Documents

32713

Supelco

Spirotetramat

PESTANAL®, analytical standard

Synonym(s):

cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl ethyl carbonate, cis-4-(Ethoxycarbonyloxy)-8-methoxy-3-(2,5-xylyl)-1-azaspiro[4.5]dec-3-en-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO5
CAS Number:
Molecular Weight:
373.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)OC1=C(C(=O)NC12CCC(CC2)OC)c3cc(C)ccc3C

InChI

1S/C21H27NO5/c1-5-26-20(24)27-18-17(16-12-13(2)6-7-14(16)3)19(23)22-21(18)10-8-15(25-4)9-11-21/h6-7,12,15H,5,8-11H2,1-4H3,(H,22,23)/t15-,21+

InChI key

CLSVJBIHYWPGQY-GGYDESQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Spirotetramat is a systemic insecticide which belongs to the class of ketoenols primarily used for the control of a wide spectrum of sucking insects. Its mode of action involves the inhibition of acetyl-CoA carboxylase enzyme activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoudou Sétamou et al.
PloS one, 12(12), e0187921-e0187921 (2017-12-14)
Huanglongbing is a devastating disease of citrus. In this study, a comprehensive profile of phloem sap amino acids (AA) in four permissive host plants of Candidatus Liberibacter asiaticus (CLas) and three non-permissive Rutaceae plants was conducted to gain a better
Lethal and sublethal effects of spirotetramat on the mealybug destroyer, Cryptolaemus montrouzieri.
Planes L, et al.
Journal of pest science, 86(2), 321-327 (2013)
David G Riley et al.
Journal of economic entomology, 112(4), 1526-1540 (2019-03-30)
Whitefly-transmitted Tomato yellow leaf curl virus (Family Geminiviridae; Genus Begomovirus) severely restricts tomato production in the Southeastern United States. Whitefly and tomato yellow leaf curl virus management studies typically investigate control tactics individually, but successful management of this pest complex
Xiao-Jun Chen et al.
Journal of AOAC International, 101(3), 848-857 (2017-10-07)
With the purpose of guaranteeing the safe use of spirotetramat and preventing its potential health threats to consumers, a QuEChERS extraction method coupled with LC triple-quadrupole tandem MS was applied in this study to determine residual spirotetramat metabolites in different
Regina K Cruzado et al.
PloS one, 15(4), e0231973-e0231973 (2020-04-29)
Potato psyllid (Bactericera cockerelli Sulc)-transmitted "Candidatus Liberibacter solanacearum" (Lso) has been negatively impacting the potato industry in the United States as well as other potato-producing countries. Lso has been linked to a condition known as zebra chip (ZC) that affects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service