Skip to Content
Merck
All Photos(2)

Key Documents

32051

Supelco

Propargite

PESTANAL®, analytical standard, technical mixture

Synonym(s):

2-(4-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O4S
CAS Number:
Molecular Weight:
350.47
Beilstein:
6934289
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(OC2CCCCC2OS(=O)OCC#C)cc1

InChI

1S/C19H26O4S/c1-5-14-21-24(20)23-18-9-7-6-8-17(18)22-16-12-10-15(11-13-16)19(2,3)4/h1,10-13,17-18H,6-9,14H2,2-4H3

InChI key

ZYHMJXZULPZUED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propargite, a non-systemic acaricide, is most commonly used on crops such as cotton, vines, nuts, tea etc for controlling phytophagous mite infestation.

Application

Propargite has been used as pesticide standard for determination of residual pesticides from:
  • Olive oil samples using GC–MS and HPLC–MS methods.
  • Tea leaf extract using GC-negative chemical ionization mass spectrometry (GC-NCI-MS) and UHPLC-tandem mass spectrometry (UHPLC-MS/MS).
  • Apple leaf extract using HPLC method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

160.5 °F - Pensky-Martens closed cup

Flash Point(C)

71.4 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dissipation behavior of propargite--an acaricide residues in soil, apple (Malus pumila) and tea (Camellia sinensis).
Kumar V et.al.
Chemosphere, 58(6), 837-843 (2005)
Analysis of agricultural residues on tea using d-SPE sample preparation with GC-NCI-MS and UHPLC-MS/MS.
Zhang X
Journal of Agricultural and Food Chemistry, 58(22), 11553-11560 (2010)
T E Archer et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 20(4), 445-456 (1985-08-01)
Residues of five different pesticides applied to alfalfa seed crops were determined in the harvested seeds and in sprouts grown from these seeds. Although seeds are usually used for future production of alfalfa plants, some of these seeds may be
Mário Eidi Sato et al.
Experimental & applied acarology, 42(2), 107-120 (2007-06-06)
The purpose of this study was to evaluate the performance of Neoseiulus californicus (McGregor) for the control of Tetranychus urticae Koch in commercial strawberry fields, under greenhouse conditions, in association or not with the use of acaricides. The N. californicus
Soumik Sarkar et al.
Journal of hazardous materials, 174(1-3), 295-298 (2009-11-07)
Biodegradation of miticide propargite was carried out in vitro by selected Pseudomonas strains isolated from tea rhizosphere. A total number of 13 strains were isolated and further screened based on their tolerance level to different concentrations of propargite. Five best

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service