Skip to Content
Merck
All Photos(2)

Key Documents

32510

Supelco

Chlorantraniliprole

PESTANAL®, analytical standard

Synonym(s):

3-Bromo-4′-chloro-1-(3-chloro-2-pyridyl)-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide, 3-Bromo-N-[4-chloro-2-methyl-6-[(methylamino)carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-1H-pyrazole-5-carboxamide, DPX E2Y45

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14BrCl2N5O2
CAS Number:
Molecular Weight:
483.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)c1cc(Cl)cc(C)c1NC(=O)c2cc(Br)nn2-c3ncccc3Cl

InChI

1S/C18H14BrCl2N5O2/c1-9-6-10(20)7-11(17(27)22-2)15(9)24-18(28)13-8-14(19)25-26(13)16-12(21)4-3-5-23-16/h3-8H,1-2H3,(H,22,27)(H,24,28)

InChI key

PSOVNZZNOMJUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorantraniliprole is a novel anthranilic diamide insecticide belonging to the class of selective ryanodine receptor agonists. It is a selective and potent activator of insect ryanodine receptors (RyRs) that are critical for muscle contraction. It is very efficient for controlling lepidopteran insect pests, as well as some species in the orders Diptera, Hemiptera, and Coleoptera.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Chlorantraniliprole activates the RyRs in insects thereby affecting calcium homeostasis by unregulated release of internal calcium in the cell, leading to feeding cessation, muscle paralysis, lethargy, and ultimately death of the insect.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Madhusmita Sahu et al.
Ecotoxicology and environmental safety, 180, 288-294 (2019-05-18)
An experiment was set up to determine the rate of dissipation of chlorantraniliprole (CTP) from two soils with contrasting properties. The other objective of the study was to find out the effect of CTP on soil microorganisms (population, microbial biomass
Jun Zhao et al.
International journal of molecular sciences, 20(5) (2019-03-03)
Uridine diphosphate glycosyltransferases (UGTs) are multifunctional detoxification enzymes, which are involved in metabolizing various chemicals and contribute to the development of insecticide resistance. However, the possible roles of UGTs in chlorantraniliprole resistance in Chilo suppressalis have rarely been studied in
Effects of chlorantraniliprole on eggs and larvae of Lobesia botrana (Denis & Schiffermuller)(Lepidoptera: Tortricidae).
Ioriatti, Claudio, et al.
Pest Management Science, 65 (6), 717-722 (2009)
Falin He et al.
Scientific reports, 9(1), 10328-10328 (2019-07-19)
Agrotis ipsilon (Lepidoptera: Noctuidae) is a major underground pest that damages many agricultural crops in China and other countries. A diet-incorporation-based bioassay was conducted to evaluate the sublethal effects of the novel anthranilic diamide chlorantraniliprole on the nutritional physiology, enzymatic
Feeding cessation effects of chlorantraniliprole, a new anthranilic diamide insecticide, in comparison with several insecticides in distinct chemical classes and mode-of-action groups.
Hannig, Greg T., Melissa Ziegler, and Paula G. Marcon.
Pest Management Science, 65 (9), 969-974 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service