Skip to Content
Merck
All Photos(2)

Documents

34174

Supelco

Pyriproxyfen

PESTANAL®, analytical standard

Synonym(s):

2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO3
CAS Number:
Molecular Weight:
321.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(COc1ccc(Oc2ccccc2)cc1)Oc3ccccn3

InChI

1S/C20H19NO3/c1-16(23-20-9-5-6-14-21-20)15-22-17-10-12-19(13-11-17)24-18-7-3-2-4-8-18/h2-14,16H,15H2,1H3

InChI key

NHDHVHZZCFYRSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyriproxifen is an insect growth regulator or pesticide, which can be used to control insects at a narrow scale. Its mode of action involves the inhibition of the process of metamorphosis of immature scale.

Application

Pyriproxifen may be used as an analytical reference standard for the determination of the analyte in gaseous and particulate phases in the atmosphere, oranges, globe artichoke leaves and fruits, water and wastewater, and different classes of food and feed by various analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beniamino Caputo et al.
PLoS neglected tropical diseases, 6(8), e1793-e1793 (2012-09-07)
The main constraint to the fight against container-breeding mosquito vectors of human arboviruses is the difficulty in targeting the multiplicity of larval sources, mostly represented by small man-made water containers. The aim of this work is to assess the feasibility
Effects of the parasitic wasp Coccobius fulvus on cycad aulacaspis scale, Aulacaspis yasumatsui, at Montgomery Botanical Center, Miami, Florida
Wiese C, et al.
Proceedings of the Florida State Horticultural Society, 118 (2005)
Determination of pesticide residues in globe artichoke leaves and fruits by GC-MS and LC-MS/MS using the same QuEChERS procedure
Machado I, et al.
Food Chemistry, 227, 227-236 (2017)
Management of cycad aulacaspis scale, Aulacaspis yasumatsui Takagi
Emshousen C, et al.
Proceedings of the Florida State Horticultural Society, 117 (2004)
Cristiane C Otoni et al.
Journal of veterinary internal medicine, 32(3), 999-1008 (2018-04-07)
Little information is available of markers that assess the disease course in dogs with idiopathic inflammatory bowel disease (IBD). Evaluate relationship between disease severity and serum and fecal biomarkers in dogs with idiopathic IBD before and after treatment. Sixteen dogs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service