Skip to Content
Merck
All Photos(1)

Documents

32407

Supelco

Cymiazole

PESTANAL®, analytical standard

Synonym(s):

2,4-Dimethyl-N-(3-methyl-2(3H)-thiazolylidene)benzenamine, 2-(2,4-Dimethylphenylimino)-3-methylthiazoline, 2-(2,4-Xylylimino)-3-methylthiazoline, N-[(2EZ)-3-Methyl-1,3-thiazol-2-(3H)ylidene]-2,4-xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2S
CAS Number:
Molecular Weight:
218.32
Beilstein:
1213211
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN1C=CSC1=Nc2ccc(C)cc2C

InChI

1S/C12H14N2S/c1-9-4-5-11(10(2)8-9)13-12-14(3)6-7-15-12/h4-8H,1-3H3/b13-12-

InChI key

YUAUPYJCVKNAEC-SEYXRHQNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cymiazole may be used as an analytical reference standard for the determination of the analyte in honey, royal jelly and beeswax by modified QuEChERS (quick, easy, cheap, effective, rugged, and safe) method coupled with LC-MS/MS (liquid chromatography tandem mass spectrometry) and solid phase extraction (SPE), followed by gas chromatography-mass spectrometry (GC-MS) analysis, respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Kryger et al.
Bulletin of entomological research, 96(6), 597-603 (2007-01-05)
In a bioassay to determine non-target ecotoxicological effects of a pyrethroid spray (Ektoban) on dung beetles, dung from both cypermethrin/cymiazol-treated and control cattle was collected one, two, three, five, seven, 14, 21 and 28 days after treatment and fed to
Development and validation of modified QuEChERS method coupled with LC--MS/MS for simultaneous determination of cymiazole, fipronil, coumaphos, fluvalinate, amitraz, and its metabolite in various types of honey and royal jelly.
Zheng W, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1072, 60-69 (2018)
Zoran Stanimirovic et al.
Mutation research, 588(2), 152-157 (2005-11-29)
Apitol, with cymiazole hydrochloride as the active ingredient, is used in bee-keeping against the ectoparasitic mite Varroa destructor. The preparation was evaluated for genotoxicity in cultured human peripheral blood lymphocytes. Sister chromatid exchange, the mitotic index and the cell proliferation
Determination of amitraz and other acaricide residues in beeswax.
Korta E, et al.
Analytica Chimica Acta, 475(1-2), 97-103 (2003)
P Cabras et al.
Journal of AOAC International, 76(1), 92-94 (1993-01-01)
A liquid chromatographic method is described for the determination of cymiazole residues in honey. This acaricide is determined on a reversed-phase (C18) column, with a CH3CN-0.001N HCl-NaCl mixture (950 mL + 50 mL + 0.3 g/L) as the mobile phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service