Skip to Content
Merck
All Photos(2)

Key Documents

43989

Supelco

Cefapirin sodium

analytical standard

Synonym(s):

3-[(Acetyloxy)methyl]-8-oxo-7-{(4-pyridinylthio)acetyl]amino}-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, Cephapirin sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N3NaO6S2
CAS Number:
Molecular Weight:
445.45
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CSc3ccncc3)C2=O)C([O-])=O

InChI

1S/C17H17N3O6S2.Na/c1-9(21)26-6-10-7-28-16-13(15(23)20(16)14(10)17(24)25)19-12(22)8-27-11-2-4-18-5-3-11;/h2-5,13,16H,6-8H2,1H3,(H,19,22)(H,24,25);/q;+1/p-1/t13-,16-;/m1./s1

InChI key

VGEOUKPOQQEQSX-OALZAMAHSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Apparao et al.
Journal of dairy science, 92(6), 2589-2597 (2009-05-19)
The selection of antimicrobial agents for the treatment of mastitis has often been based on results of in vitro susceptibility testing. However, the results of in vitro susceptibility tests have been shown to be poor predictors of treatment outcomes. The
Abd el-Gawad Salah el-Tahawy et al.
Research in veterinary science, 90(1), 44-50 (2010-05-15)
A total of 412 multiparous German Holstein cows were screened for postpartum pyometra, follicular cysts and ovarian inactivity to assess economic and productivity losses in relation to pharmaceutical expenditures. Our results show that cows treated for pyometra with prostaglandin f2
J Kneebone et al.
Journal of dairy science, 93(9), 3961-3964 (2010-08-21)
Rapid antibiotic screening tests are widely used in the dairy industry to monitor milk for the presence of antibiotic residues above regulated levels. Given the persistent concern over contamination of milk products with antibiotic residues, we investigated the utility of
R M Stockler et al.
Journal of dairy science, 92(9), 4262-4275 (2009-08-25)
The objective was to determine the effect of milking frequency and dosing interval on pharmacokinetics of cephapirin after intramammary infusion. Six healthy Holstein cows were administered cephapirin (200 mg) into 1 rear mammary gland after each of 2 milkings. Cows
Jean-Philippe Roy et al.
The Canadian veterinary journal = La revue veterinaire canadienne, 50(12), 1257-1262 (2010-03-02)
This study determined the efficacy of a 5-day extended therapy with cephapirin sodium in dairy cows chronically infected with Staphylococcus aureus. Chronically infected cows selected from 14 dairy herds in the St-Hyacinthe region, Québec were randomly allocated to a group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service