Skip to Content
Merck
All Photos(1)

Documents

PHR1798

Supelco

Levamisole Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
Beilstein:
4358988
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 212
traceable to Ph. Eur. L0380000
traceable to USP 1359302

API family

levamisole

CofA

current certificate can be downloaded

packaging

pkg of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

228-230 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.C1CN2C[C@@H](N=C2S1)c3ccccc3

InChI

1S/C11H12N2S.ClH/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10;/h1-5,10H,6-8H2;1H/t10-;/m1./s1

InChI key

LAZPBGZRMVRFKY-HNCPQSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Levamisole Hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by potentiometric sensing, spectrometric and atomic absorption spectroscopy methods.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3172 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indirect determination of levamisole by AAS and precipitation in a continuous-flow assembly
Ortiz SL, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 48(1), 112-117 (1993)
Potentiometric sensing of levamisole hydrochloride based on molecularly imprinted polymer
Sadeghi S, et al.
Sensors and Actuators B, Chemical, 122(1), 158-164 (2007)
Sreeparna Pradhan et al.
Current biology : CB, 29(17), 2867-2879 (2019-08-20)
Foraging strategies should be tuned to the expected distribution of resources in the environment. Tuning can occur over generations and lead to genetic differences in innate foraging behavior or over shorter timescales within an individual's lifespan. Both genetically encoded and
Jote T Bulcha et al.
Cell reports, 26(2), 460-468 (2019-01-10)
Biological systems must possess mechanisms that prevent inappropriate responses to spurious environmental inputs. Caenorhabditis elegans has two breakdown pathways for the short-chain fatty acid propionate: a canonical, vitamin B12-dependent pathway and a propionate shunt that is used when vitamin B12
Davide Caprini et al.
Advanced biology, 5(9), e2100927-e2100927 (2021-08-24)
AWC olfactory neurons are fundamental for chemotaxis toward volatile attractants in Caenorhabditis elegans. Here, it is shown that AWCON responds not only to chemicals but also to mechanical stimuli caused by fluid flow changes in a microfluidic device. The dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service