Skip to Content
Merck
All Photos(2)

Key Documents

31400

Millipore

Dextrin from potato starch

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H12O6)x
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

form

powder

Quality Level

application(s)

microbiology

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

Dextrin is basically a sweet sugar extracted from potato starch. Dextrin color range is from white to tan; it has low to high solubilities in cold water, and gives pastes that vary widely in viscosity. It is produced from all commercial grain and tuber starches. During its manufacture, the factors taken into consideration are type of starch and moisture content, roasting time and temperature, and the type and amount of catalyst used.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Starch: Chemistry and Technology.
Whistler RL
Technology & Engineering, 595-596 (2012)
The Organic Gardener's Handbook of Natural Insect and Disease Control.
Ellis BW
Science, 484-485 (1996)
C Costalos et al.
Acta paediatrica Scandinavica, 74(3), 382-385 (1985-05-01)
Serial plasma glucose estimations were performed in 44 light-for-date infants and 17 infants of diabetic mothers fed 2, 3 or 4-hourly with feeds containing 10% additional carbohydrate in the form of a glucose polymer (Caloreen). In the infants fed 2
G Russell et al.
European journal of pediatrics, 143(1), 32-34 (1984-11-01)
Stool composition was studied in 10 newborn infants following (a) a modified milk formula (Cow & Gate Premium) and (b) a high energy formula [104 kcal (435 kJ) per 100 ml] in which the additional energy was provided in the
The beginning of icodextrin.
Chandra D Mistry
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis, 31 Suppl 2, S49-S52 (2011-09-29)

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service