Skip to Content
Merck
All Photos(3)

Key Documents

419699

Sigma-Aldrich

Maltodextrin

Synonym(s):

MLD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

biological source

corn kernels

description

dextrose equivalent 16.5 - 19.5

form

powder

mp

240 °C (dec.) (lit.)

SMILES string

O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1

InChI key

GZCGUPFRVQAUEE-SLPGGIOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltodextrin is a saccharide polymer that can be classified as a carbohydrate. It can be produced by enzymatic or acid hydrolysis of the starch. The material formed after purification and spray drying can be used in a variety of food and beverage products. It can be used as a good source of energy (16 kJ/g) in food products.

Application

  • Formulation and characterization of catechin-loaded proniosomes for food fortification.: This study investigates the use of maltodextrin in the formulation of catechin-loaded proniosomes, highlighting its potential in food fortification applications. The results suggest significant benefits in terms of stability and bioavailability of catechins when encapsulated in proniosomes using maltodextrin (Shruthi et al., 2021).
  • Impact of excipient choice on the aerodynamic performance of inhalable spray-freeze-dried powders.: This research explores how different excipients, including maltodextrin, affect the aerodynamic performance of spray-freeze-dried powders for inhalation. The findings emphasize the critical role of excipient selection in optimizing drug delivery efficiency (Wanning et al., 2020).
  • Influence of spray drying on the stability of food-grade solid lipid nanoparticles.: The study examines the impact of spray drying on the stability of solid lipid nanoparticles, with maltodextrin serving as a stabilizer. The results indicate that maltodextrin effectively enhances the stability and shelf life of these nanoparticles in food applications (Salminen et al., 2019).
  • The effect of binary mixture composition and magnesium stearate concentration on the Hiestand Tableting Indices and other related mechanical properties.: This article investigates the mechanical properties of tablet formulations, focusing on the role of maltodextrin in binary mixtures. The study provides insights into the optimization of tablet formulations for improved mechanical strength and performance (Likitlersuang et al., 2007).
  • The 3D model: explaining densification and deformation mechanisms by using 3D parameter plots.: This paper presents a comprehensive model to explain the densification and deformation mechanisms in tablet formulations, using maltodextrin as a case study. The 3D parameter plots offer a detailed understanding of the compaction behavior of pharmaceutical excipients (Picker, 2004).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nutrition, health, and regulatory aspects of digestible maltodextrins
Hofman DL, et al.
Critical Reviews in Food Science and Nutrition, 56(12), 2091-2100 (2016)
Effect of addition of maltodextrin on drying kinetics and stickiness of sugar and acid-rich foods during convective drying: experiments and modelling
Adhikari B, et al.
Journal of Food Engineering, 62(1), 53-68 (2004)
Geoffrey Livesey et al.
The American journal of clinical nutrition, 89(1), 114-125 (2009-01-08)
The glycemic response to diet has been linked with noncommunicable diseases and is reduced by low-palatable, viscous, soluble fiber (1). Whether a palatable, low-viscous, soluble fiber such as resistant maltodextrin (RMD) has the same effect is unclear. The objective was
David J Baer et al.
The Journal of nutrition, 144(7), 1023-1029 (2014-04-20)
Resistant maltodextrin (RM) is a novel soluble, nonviscous dietary fiber. Its metabolizable energy (ME) and net energy (NE) values derived from nutrient balance studies are unknown, as is the effect of RM on fecal microbiota. A randomized, placebo-controlled, double-blind crossover
Pedro B Júdice et al.
Applied physiology, nutrition, and metabolism = Physiologie appliquee, nutrition et metabolisme, 38(1), 49-56 (2013-02-02)
Research on the effect of caffeine on energy expenditure (EE), physical activity (PA), and total sleep time (TST) during free-living conditions using objective measures is scarce. We aimed to determine the impact of a moderate dose of caffeine on TST

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service