Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dextrin from potato starch
Linear Formula:
(C
6
H
12
O
6
)
x
CAS Number:
9004-53-9
EC Number:
232-675-4
Products
Product Category
carbohydrates (2)
Brand
Millipore (2)
Gene Alias
Physical Form
Quality Segment
Application
Special Grade
Biotechnology (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
31400
Dextrin from potato starch,
suitable for microbiology
Expand
View Pricing
31405
Dextrin from potato starch,
for biotechnological purposes
Expand
View Pricing