Skip to Content
Merck
All Photos(2)

Key Documents

A8515

Sigma-Aldrich

Amylopectin from potato starch

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

potato

form

powder

impurities

<25% water (Karl Fischer)

solubility

water: 7.69 mg/mL, clear to hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O[C@H]5O[C@@H]([C@H]([C@@H]([C@H]5O)O)O)CO)CO)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C30H52O26/c31-1-6-11(35)13(37)19(43)28(50-6)55-24-9(4-34)52-27(21(45)16(24)40)48-5-10-25(56-29-20(44)14(38)12(36)7(2-32)51-29)17(41)22(46)30(53-10)54-23-8(3-33)49-26(47)18(42)15(23)39/h6-47H,1-5H2/t6-,7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26+,27+,28-,29-,30-/m1/s1

InChI key

WMGFVAGNIYUEEP-WUYNJSITSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amylopectin is a glucose-based, highly-branched polysaccharide which together with amylose makes starch. The glucosidic bonds of amylopectin are α-1,4 in the glucan chain and α-1,6 at the branch points. Amylopectin from maize differs in structure form amylopectin from other plant species and is used in the study of the development of starches.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sushil Dhital et al.
Journal of agricultural and food chemistry, 59(18), 10151-10161 (2011-08-16)
Chemical composition, molecular structure and organization, and thermal and pasting properties of maize and potato starches fractionated on the basis of granule size were investigated to understand heterogeneity within granule populations. For both starches, lipid, protein, and mineral contents decreased
Shazia Juna et al.
Journal of chromatography. A, 1219, 161-172 (2011-12-07)
A detailed study of the elution behaviour, apparent molecular characteristics and hydrodynamic properties of amylopectin-type fraction (isolated from normal corn starch) in aqueous media employing asymmetrical flow field-flow fractionation (AF4) was undertaken by systematically varying the channel flow (F(ch)), cross
Shujun Wang et al.
Scientific reports, 6, 20965-20965 (2016-02-11)
Starch retrogradation is a term used to define the process in which gelatinized starch undergoes a disorder-to-order transition. A thorough understanding of starch retrogradation behavior plays an important role in maintaining the quality of starchy foods during storage. By means
Anas Iqbal et al.
Saudi journal of biological sciences, 28(2), 1283-1296 (2021-02-23)
Improving grain filling in the presernt farming systems is crucial where grain filling is a concern due to the extreme use of chemical fertilizers (CF). A field experiment was conducted at the experimental station of Guangxi University, China in 2019
Varatharajan Vamadevan et al.
Carbohydrate polymers, 92(2), 1653-1659 (2013-02-13)
The relationship between the internal structure of amylopectin from diverse plants and thermal properties of the starch granules has been investigated. Correlations were found between structural parameters, such as number of building blocks in clusters, interblock chain length and length

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service