Skip to Content
Merck
All Photos(1)

Key Documents

22199

Supelco

Cellulose triacetate

Selectophore

Synonym(s):

Triacetylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

granular

solubility

chloroform: 0.1 g/10 mL, clear, colorless (leave for 5 h)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. P. Mishra
A Text Book of Fibre Science and Technology, 187-187 (2000)
Fabrication and positron annihilation spectroscopy (PAS) characterization of cellulose triacetate membranes for forward osmosis.
Ong, Rui Chin, and Tai-Shung Chung
Journal of Membrane Science, 394, 230-240 (2012)
Facilitated transport of small carbohydrates through plasticized cellulose triacetate membranes. Evidence for fixed-site jumping transport mechanism.
Riggs, Jennifer A., and Bradley D. Smith.
Journal of the American Chemical Society, 119, 2765-2766 (1997)
Asma Yahia Cherif et al.
Journal of hazardous materials, 227-228, 386-393 (2012-06-15)
In this work, we have developed a novel class of polymeric inclusion membranes (PIMs) for the cations separation. The membrane is made up of cellulose triacetate modified by poly-electrolytes (poly-phosphoric acid, polyvinyl pyrolidone, polyacrylic acid, polyvinyl alcohol and poly-anetholsulfonic acid)
Shuangxin Liu et al.
Blood purification, 29(4), 375-382 (2010-05-21)
Thrombocytopenia is a common complication in patients receiving continuous veno-venous hemofiltration (CVVH) in the intensive care unit. The hemofilter itself plays an important role in the genesis of thrombocytopenia. The present study was undertaken to test whether there were differences

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service