Skip to Content
Merck
All Photos(1)

Key Documents

I3380

Supelco

Iodine

ReagentPlus®, ≥99.8% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050601
PubChem Substance ID:
NACRES:
NA.21

vapor density

9 (vs air)

Quality Level

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

product line

ReagentPlus®

Assay

≥99.8% (titration)

resistivity

1.3E15 μΩ-cm

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Iodine-Promoted Reductive Sulfenylation Using Ketones as Hydride Donors: Describes a novel iodine-mediated synthesis method, expanding the toolkit for chemical synthesis with potential applications in pharmaceutical research (Duan Y et al., 2024).

  • Phenyliodine bis(trifluoroacetate) as a sustainable reagent: Discusses the use of iodine compounds in sustainable chemistry, emphasizing their importance in organic synthesis and potential for greener chemical processes (Kumar S et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tim I M Korevaar et al.
The Journal of clinical endocrinology and metabolism, 98(11), 4382-4390 (2013-09-17)
Premature delivery is an important risk factor for child mortality and psychiatric, metabolic, and cardiovascular disease later in life. In the majority of cases, the cause of prematurity cannot be identified. Currently, it remains controversial whether abnormal maternal thyroid function
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and
Shao J Zhou et al.
The American journal of clinical nutrition, 98(5), 1241-1254 (2013-09-13)
Routine iodine supplementation during pregnancy is recommended by leading health authorities worldwide, even in countries where the iodine status of the population is sufficient. We evaluated the efficacy and safety of iodine supplementation during pregnancy or the periconceptional period on
Elizabeth N Pearce et al.
Thyroid : official journal of the American Thyroid Association, 23(5), 523-528 (2013-03-12)
Dietary iodine intake is required for the production of thyroid hormone. Consequences of iodine deficiency include goiter, intellectual impairments, growth retardation, neonatal hypothyroidism, and increased pregnancy loss and infant mortality. In 1990, the United Nations World Summit for Children established
Michael B Zimmermann
Thyroid : official journal of the American Thyroid Association, 17(9), 829-835 (2007-10-25)
Iodine is required for the production of thyroid hormones, which are essential for normal brain development, and the fetus, newborn, and young child are particularly vulnerable to iodine deficiency. The iodine requirement increases during pregnancy and recommended intakes are in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service