Skip to Content
Merck
All Photos(3)

Key Documents

181315

Sigma-Aldrich

Polyacrylonitrile

average Mv 150,000

Synonym(s):

Vinyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H3N)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mv 150,000

refractive index

n20/D 1.514

transition temp

Tm 317 °C
Tg 85 °C

density

1.184 g/mL at 25 °C (lit.)

SMILES string

C=CC#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PAN molecule has strong polar nitrile groups.It is relatively insoluble in nature. PAN based carbon fibers possess very high strength compared to other polymeric precursors. When subjected to heat treatment, it can produce high carbon yield giving rise to thermally stable, highly oriented, graphite like molecular structure. Generating carbon-fiber from PAN based fiber is a combination of three processes-- namely stabilization, carbonization and graphitization.

Application

Polyacrylonitrile (PAN) is used as polymeric carbon precursor to form carbon fibers, electrospun activated carbon materials having meso-macro pores, carbon black additives.These are consecutively used in hydrogen storage, EMI shielding, electrochemistry, separation processes. It may find applications in PAN based single walled carbon nanotube composites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Sun Im et al.
Journal of colloid and interface science, 314(1), 32-37 (2007-06-19)
Mesoporous activated carbon samples were prepared from electrospun PAN-based carbon fibers using physical activation with silica. Textural characterization was performed using nitrogen adsorption at 77 K. The BET specific surface area and pore size distribution of silica activated carbon materials
Fluorination effects of carbon black additives for electrical properties and EMI shielding efficiency by improved dispersion and adhesion.
Im JS, et al.
Carbon, 47(11), 2640-2647 (2009)
A review of heat treatment on polyacrylonitrile fiber.
Rahaman MS, et al.
Polymer Degradation and Stability, 92(8), 1421-1432 (2007)
Polyacrylonitrile single-walled carbon nanotube composite fibers.
Sreekumar TV, et al.
Advanced Materials, 16(1), 58-61 (2004)
Prevention of dialysis-related amyloidosis using the polyacrylonitrile AN69 membrane.
J Chanard
Contributions to nephrology, 112, 137-144 (1995-01-01)

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service