Skip to Content
Merck
All Photos(2)

Key Documents

22150

Millipore

D-(+)-Cellobiose

≥99.0%, suitable for microbiology, Reducing sugar disaccharide

Synonym(s):

Cellobiose, Cellose, Beta-Cellobiose, D-Cellobiose, β-D-Glc-(1→4)-D-Glc, 4-O-β-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93795
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +34±1°, 15 hr, c = 10% in H2O

mol wt

342.30 g/mol

ign. residue

≤0.1% (as SO4)

loss

≤1% loss on drying, 110 °C

mp

~235 °C (dec.)
239 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤150 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12-/m0/s1

InChI key

DKXNBNKWCZZMJT-WELRSGGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellobiose is a disaccharide that is a reducing sugar. It is added to culture media as a nutrient supplement, specifically as a carbohydrate source, and can be used to differentiate bacteria based on their ability to ferment cellobiose. For example, Vibrio spp. can ferment cellobiose which results in acid production and can be indicated by the pH indicator bromothymol blue, which turns yellow at acidic pH. The addition of cellobiose in the media also stimulates the initial growth of Salmonella.

Application

Cellobiose has applications as a carbohydrate source in culture media and biochemical characterization of microorganisms, based on their carbohydrate fermentation abilities

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corinna Schiano-di-Cola et al.
The FEBS journal, 287(12), 2577-2596 (2019-11-23)
Thermostable cellulases from glycoside hydrolase family 7 (GH7) are the main components of enzymatic mixtures for industrial saccharification of lignocellulose. Activity improvement of these enzymes via rational design is a promising strategy to alleviate the industrial costs, but it requires
Benjamin J Matthews et al.
eLife, 8 (2019-05-22)
Female Aedes aegypti mosquitoes are deadly vectors of arboviral pathogens and breed in containers of freshwater associated with human habitation. Because high salinity is lethal to offspring, correctly evaluating water purity is a crucial parenting decision. We found that the
Alessandro Boianelli et al.
PloS one, 7(10), e47393-e47393 (2012-10-31)
When grown on glucose and beta-glucosides, S. pneumoniae shows sequential use of sugars resulting in diauxic growth with variable time extent of the lag phase separating the biphasic growth curve. The pneumococcal beta-glucoside uptake locus containing the PTS transporter spr0276-82
Lara Hassan et al.
mBio, 10(4) (2019-07-04)
It is essential for microbes to acquire information about their environment. Fungi use soluble degradation products of plant cell wall components to understand the substrate composition they grow on. Individual perception pathways have been well described. However, the interconnections between
Hoi Yee Keung et al.
Applied and environmental microbiology, 83(7) (2017-01-25)
Bifidobacteria exert beneficial effects on hosts and are extensively used as probiotics. However, due to the genetic inaccessibility of these bacteria, little is known about their mechanisms of carbohydrate utilization and regulation. Bifidobacterium breve strain JCM1192 can grow on water-insoluble

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service