Skip to Content
Merck
All Photos(2)

Documents

D0256

Sigma-Aldrich

Dulcitol

≥99% (GC)

Synonym(s):

Dulcite, Galactitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721903
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (GC)

form

crystals

sweetness

<0.6 × sucrose

color

white

solubility

water: 50 mg/mL, clear, colorless

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5+,6-

InChI key

FBPFZTCFMRRESA-GUCUJZIJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dulcitol (Galactitol), a sugar alcohol derived through reduction of galactose, is important in studies related to cataract formation, hepatosplenomegaly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). Galactitol may be used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

Biochem/physiol Actions

A sugar alcohol and slightly sweet tastant for human taste cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukas Müller et al.
Materials (Basel, Switzerland), 13(20) (2020-10-15)
Measuring thermo-physical properties of phase change materials (PCM) in a consistent and reliable manner is essential for system layout of thermal energy storages and correspondingly material selection. Only if basic properties are assessed in a comparable way a selection process
Sandra K Parsons et al.
Applied and environmental microbiology, 81(17), 5804-5811 (2015-06-21)
The life cycles of many enteric bacterial species require a transition between two very distinct environments. Their primary habitat is the gastrointestinal tract of the host, while their secondary habitat, during transmission from one host to another, consists of environments
Ignasi Barba et al.
Investigative ophthalmology & visual science, 51(9), 4416-4421 (2010-04-09)
To explore the metabolic profile of vitreous fluid of patients with proliferative diabetic retinopathy (PDR) using 1H-NMR-based metabonomic analysis. 1H-NMR spectra were acquired from vitreous samples obtained during vitrectomy from 22 patients with type 1 diabetes with PDR and from
H P Schwarz et al.
Clinical chemistry, 31(3), 420-422 (1985-03-01)
An oral dose of galactose, 1 g/kg of body weight, was administered to 24 children with the Duarte variant/classical galactosemia genetic compound (GtD/gt) and to 16 controls ranging in age from 0.3 to 10.7 years. Urine was then collected for
G T Berry et al.
The Journal of pediatrics, 138(2), 260-262 (2001-02-15)
In a newborn infant with galactose-1-phosphate uridyltransferase deficiency and encephalopathy, brain magnetic resonance imaging revealed cytotoxic edema in white matter. Using in vivo proton magnetic resonance spectroscopy, we detected approximately 8 mmol galactitol per kilogram of brain tissue, an amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service