Skip to Content
Merck
All Photos(3)

Key Documents

M5895

Sigma-Aldrich

D-(+)-Maltose monohydrate

≥98% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

potato

Quality Level

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

optical activity

[α]20/D 128.0 to 132.0 °, c = 4% (w/v) in water + trace NH4OH

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

impurities

≤0.3% glucose
≤1.0% maltotriose

color

white

useful pH range

5.0-7 (25 °C, 180 g/L)

mp

102-103  °C

solubility

H2O: 50 mg/mL

application(s)

agriculture

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Maltose monohydrate is a disaccharide sugar made up of two α-d-glucose molecules joined by α(1→4) glycosidic bond.[1] Apparent molar volumes of D-(+)-maltose monohydrate has been studied in water and guanidine hydrochloride solutions by density measurement experiment.[1]

Application

D-(+)-Maltose monohydrate is the suitable reagent for the following studies:
  • Cell culture studies.
  • Insect cell culture studies.
  • To study the glass transition temperatures and phase relations for several saccharide-water systems.[2]
  • As medium supplement in the modified Luria-Bertani broth (LBM) medium for E. coli strains.[3]
It may be employed as standard for the α-amylase and invertase assays.[4]

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fusé et al.
Peptides, 20(11), 1285-1293 (1999-12-28)
Neuropeptides of the cockroach allatostatin (AST) family are known for their ability to inhibit the production of juvenile hormone by the corpora allata of cockroaches. Since their discovery, they have also been shown to modulate myotropic activity in a range
Phase relations and vitrification in saccharide-water solutions and the trehalose anomaly.
Green JL and Angell CA.
The Journal of Physical Chemistry, 93(8), 2880-2882 (1989)
Partial molar volumes of transfer of some disaccharides from water to aqueous guanidine hydrochloride solutions at 298.15 K.
Banipal TS, et al.
Indian J. Chem. A, 41(6), 1131-1138 (2002)
Roberto Vincis et al.
Current biology : CB, 30(10), 1834-1844 (2020-04-04)
Research over the past decade has established the gustatory insular cortex (GC) as a model for studying how primary sensory cortices integrate sensory, affective, and cognitive signals. This integration occurs through time-varying patterns of neural activity. Selective silencing of GC
Amar Thaker et al.
Biotechnology and bioengineering, 116(12), 3160-3167 (2019-09-11)
The use of X-ray radiation in radiotherapy is a common treatment for many cancers. Despite several scientific advances, determination of radiation delivered to the patient remains a challenge due to the inherent limitations of existing dosimeters including fabrication and operation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service