Skip to Content
Merck
All Photos(1)

Key Documents

W222100

Sigma-Aldrich

Butyric acid

≥99%, FG

Synonym(s):

Butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOH
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2221
Beilstein:
906770
EC Number:
Council of Europe no.:
5
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.005
NACRES:
NA.21
Organoleptic:
cheese; buttery; fruity
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

3.04 (vs air)

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

824 °F

expl. lim.

10 %

refractive index

n20/D 1.398 (lit.)

pH

2 (25 °C, 10 g/L)
3 (20 °C, 10 g/L)

bp

162 °C (lit.)

mp

−6-−3 °C (lit.)

density

0.964 g/mL at 25 °C (lit.)

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
Pb: ≤10 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cheese; buttery; fruity

SMILES string

CCCC(O)=O

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

FERIUCNNQQJTOY-UHFFFAOYSA-N

Gene Information

human ... HDAC1(3065)

Looking for similar products? Visit Product Comparison Guide

General description

Butyric acid is a volatile fatty acid[1] that can be used as a flavoring agent.[1] It has fruity or cheesy flavor and is mainly found in dairy products.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 359-360 (1997)
Effect of fat content on odor intensity of three aroma compounds in model emulsions: ?-decalactone, diacetyl, and butyric acid.
Guyot C, et al.
Journal of Agricultural and Food Chemistry, 44(8), 2341-2348 (1996)
Gili Berkovitch et al.
Journal of medicinal chemistry, 51(23), 7356-7369 (2008-11-15)
Multifunctional acyloxyalkyl ester prodrugs of 5-aminolevulinic acid in cancer cell lines inhibited the proteasome and induced apoptosis and heme synthesis. The most potent prodrug was butyryloxymethyl 5-amino-4-oxopentanoate (1a). The metabolically released formaldehyde from the prodrugs was the dominant factor affecting
Carlo F M Welters et al.
Diseases of the colon and rectum, 45(5), 621-627 (2002-05-11)
Inflammation is a constant finding in the ileal reservoir of patients with an ileal pouch-anal anastomosis and is associated with decreased fecal concentrations of the short chain fatty acid butyrate, increased fecal pH, changes in fecal flora, and increased concentrations
Shomik Sengupta et al.
Journal of gastroenterology and hepatology, 21(1 Pt 2), 209-218 (2006-02-08)
Butyrate, the four-carbon fatty acid, is formed in the human colon by bacterial fermentation of carbohydrates (including dietary fiber), and putatively suppresses colorectal cancer (CRC). Butyrate has diverse and apparently paradoxical effects on cellular proliferation, apoptosis and differentiation that may

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service