Skip to Content
Merck
All Photos(1)

Documents

11880

Supelco

Bathophenanthroline

for spectrophotometric det. of Fe in serum, ≥99.0%

Synonym(s):

4,7-Diphenyl-1,10-phenanthroline, BPhen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2
CAS Number:
Molecular Weight:
332.40
Beilstein:
261048
EC Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (NT)
≥99.0%

form

solid

quality

for spectrophotometric det. of Fe in serum

technique(s)

UV/Vis spectroscopy: suitable

mp

218-220 °C (lit.)
218-222 °C

SMILES string

c1ccc(cc1)-c2ccnc3c2ccc4c(ccnc34)-c5ccccc5

InChI

1S/C24H16N2/c1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18/h1-16H

InChI key

DHDHJYNTEFLIHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bathophenanthroline (BPhen) is a chelating agent and is specific for ferrous ions.

Application

Bathophenanthroline was used in an ultra-sensitive and selective nonextractive quenchofluorimetric method, in order to determine palladium (II) at μg/l Levels. It may also be used as the buffer layer to improve the performance of organic photovoltaic cells.

Other Notes

For the determination of iron in serum, in urine; for the determination of iron(II) in presence of iron(III); for the determination of total iron.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.E. Leon et al.
Analytical Chemistry, 53, 706-706 (1981)
M.J. Seven, R.E. Peterson
Analytical Chemistry, 30, 2016-2016 (1958)
R.D. Perry et al.
Analyst, 102, 114-114 (1977)
T Mizuno
Talanta, 19(3), 369-372 (1972-03-01)
Traces of iron(II) (1-30 ppM) in the presence of iron(III) were determined (error <10%) by the bathophenanthroline method. Interference of iron(III) was eliminated by masking with sodium pyrophosphate (2.5-60 mg). The iron(II) complex was extracted with n-butanol, at pH 4.2-4.7.
Organic photovoltaic cells with improved performance using bathophenanthroline as a buffer layer.
Wang N
Chinese Journal of Chemical Physics, 23 (1), 84-88 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service