Skip to Content
Merck
All Photos(1)

Documents

82940

Supelco

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt

for spectrophotometric det. of Fe, ≥99.0%

Synonym(s):

5,5′-[3-(2-Pyridyl)-1,2,4-triazine-5,6-diyl]difuran-2-sulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N4Na2O8S2
CAS Number:
Molecular Weight:
494.37
Beilstein:
5710932
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (TLC)
≥99.0%

form

crystals

quality

for spectrophotometric det. of Fe

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc(o1)-c2nnc(nc2-c3ccc(o3)S([O-])(=O)=O)-c4ccccn4

InChI

1S/C16H10N4O8S2.2Na/c21-29(22,23)12-6-4-10(27-12)14-15(11-5-7-13(28-11)30(24,25)26)19-20-16(18-14)9-3-1-2-8-17-9;;/h1-8H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2

InChI key

IVLSEFOVPQFJBB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt also known as Ferene is a chelating agent which usually binds with Fe2+ ion to form coloured complex.

Application

It was used in chromogen screening while studying non-transferrin-bound iron quantification. It was used in measuring the chelating activity on ferrous ions.

Other Notes

Colorimetric reagent for iron, which is more sensitive than other reagents (e.g. Ferrozine), molar absorptivity 35500 liters/mol cm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.J. Hennessy et al.
Canadian Journal of Chemistry, 62, 721-721 (1984)
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 28, 275-275 (1983)
Non-transferrin-bound iron assay system utilizing a conventional automated analyzer.
Ito, Satoshi, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 437, 129-135 (2014)
In vitro antioxidant, anticholinesterase and antimicrobial activity studies on three Agaricus species with fatty acid compositions and iron contents: A comparative study on the three most edible mushrooms.
Ozturk, Mehmet, et al.
Food And Chemical Toxicology, 49.6, 1353-1360 (2011)
Interaction between bovine control materials and the ferene iron method.
H M Barbour et al.
Annals of clinical biochemistry, 30 ( Pt 2), 221-222 (1993-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service