Skip to Content
Merck
All Photos(2)

Key Documents

06279

Supelco

Mangiferin

analytical standard

Synonym(s):

1,3,6,7-Tetrahydroxyxanthone C2-β-D-glucoside, Chinomin, Chinonin, Shamimin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O11
CAS Number:
Molecular Weight:
422.34
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3Oc4cc(O)c(O)cc4C(=O)c3c2O

InChI

1S/C19H18O11/c20-4-11-15(25)17(27)18(28)19(30-11)12-8(23)3-10-13(16(12)26)14(24)5-1-6(21)7(22)2-9(5)29-10/h1-3,11,15,17-23,25-28H,4H2/t11-,15-,17+,18-,19+/m1/s1

InChI key

AEDDIBAIWPIIBD-ZJKJAXBQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mangiferin is a xanthone glucoside and a naturally occurring polyphenol found abundantly in the plants of genus Mangifera particularly Mangifera indica L. and also in some medicinal herbs. Mangiferin exhibits antioxidant, antiviral, antitumor, anti-inflammatory and gene-regulatory effects and is considered to be of high research interest due to its therapeutic properties.

Application

Mangiferin may be used as an analytical standard for the determination of the analyte in rat plasma, traditional Chinese medicines and honeybush tea (Cyclopia spp.) by chromatography based techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: hypericum

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Han et al.
Journal of pharmaceutical and biomedical analysis, 51(1), 260-263 (2009-08-25)
An ultra-performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS) method employing electrospray ionization (ESI) has been developed for the determination of mangiferin in rat plasma using diphenhydramine as the internal standard (IS). Liquid-liquid extraction (LLE) was used for sample preparation and
High-performance liquid chromatographic method for the determination of mangiferin, likviritin and dihydroquercetin in rat plasma and urine
Geodakyan SV, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 577(2), 371-375 (1992)
Ronghua Dai et al.
Journal of chromatographic science, 42(4), 207-210 (2004-05-25)
High-performance liquid chromatography is employed to determine the contents of six marker components such as mangiferin, jateorrhizine, palmatine, berberine, cinnamic acid, and cinnamaldehyde in the traditional Chinese medicinal preparation Zi-Shen pill. The separation is performed on a C(18) column by
S J Lin et al.
Journal of chromatography. A, 730(1-2), 17-23 (1996-04-12)
High-performance liquid chromatography was employed to determine the contents of several marker substances such as gentiopicroside, mangiferin, palmatine, berberine, baicalin, wogonin and glycyrrhizin in Sann-Joong-Kuey-Jian-Tang. The separation was performed on a Cosmosil 5C18-AR column by gradient elution with 0.03% (v/v)
Reversed-phase HPLC determination of mangiferin, isomangiferin and hesperidin in Cyclopia and the effect of harvesting date on the phenolic composition of C. genistoides
Joubert E, et al.
European Food Research and Technology, 216(3), 270-273 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service