Skip to Content
Merck
All Photos(2)

Key Documents

18692

Supelco

Lupeol

analytical standard

Synonym(s):

β-Viscol, 20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupene, Clerodol, Fagarasterol, Lupenol, Monogynol B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H]C12CCC3C4(C)CCC(O)C(C)(C)[C@]4([H])CCC3(C)[C@]1(C)CCC5(C)CC[C@H](C(C)=C)[C@]25[H]

InChI

1S/C30H50O/c1-19(2)20-11-14-27(5)17-18-29(7)21(25(20)27)9-10-23-28(6)15-13-24(31)26(3,4)22(28)12-16-30(23,29)8/h20-25,31H,1,9-18H2,2-8H3/t20-,21+,22-,23+,24-,25+,27+,28-,29+,30+/m0/s1

InChI key

MQYXUWHLBZFQQO-QGTGJCAVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lupeol is a novel anti-cancer and anti-inflammatory dietary triterpene. They help in stabilizing phospholipid bilayers in plant cell membranes. In general, this group contains 28-29 carbons along with carbon-carbon double bonds, usually one in the sterol nucleus and at times a second in the alkyl side chain.

Application

It was used as reference standard in method development and validation of lupeol from Vernonia Cinerea using HPLC method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza sambucus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Saleem
Cancer letters, 285(2), 109-115 (2009-05-26)
In the Western world, an average of 250 mg per day of triterpenes (member of phytosterol family), largely derived from vegetable oils, cereals, fruits and vegetables is consumed by humans. During the last decade, there has been an unprecedented escalation
Ardiansyah et al.
Bioscience, biotechnology, and biochemistry, 76(1), 183-185 (2012-01-11)
Supplementation with lupeol (0.67 g·kg(-1)) of the AIN-93M-based diet fed for 7 weeks to stroke-prone spontaneously hypertensive rats caused significantly decreased blood pressure as compared with a control group. Urinary 8-hydroxy-2'-deoxyguanosine was significantly lower in the lupeol group. Finally, lupeol
Areej Mohammad Al-Taweel et al.
Molecules (Basel, Switzerland), 17(3), 2675-2682 (2012-03-07)
Nine compounds have been isolated for the first time from Celtis africana, namely trans-N-coumaroyltyramine (1), trans-N-feruloyltyramine (2), trans-N-caffeoyltyramine (3), lauric acid (4), oleic acid (5), palmitic acid (6), lupeol (7), β-sitosterol (8) and oleanolic acid (9), respectively. Their structures have
Borhane Annabi et al.
Inflammopharmacology, 21(1), 91-99 (2012-06-19)
Local inflammation-induced extracellular matrix structural changes are a prerequisite to neoplastic invasion by pediatric intracranial tumors. Accordingly, increased expression of matrix metalloproteinases MMP-2 and MMP-9, two inflammation-induced matrix metalloproteinases (MMPs), may further aid the transformed cells either to infiltrate adjacent
Development and validation of High Performance Liquid Chromatographic Method for the Simultaneous Determination of ?-sitosterol and Lupeol in Veronica cinerea Linn.
Shah, Willy, M. B. Kekare, and Vikas Vaidya.
International Journal of Pharma and Bio Sciences, 1 (3), 1-5 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service