Skip to Content
Merck
All Photos(2)

Documents

C4582

Sigma-Aldrich

β-Carotene

synthetic, ≥95% (HPLC), crystalline

Synonym(s):

β,β-Carotene, Provitamin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Beilstein:
1917416
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

crystalline

impurities

α-carotene, essentially free

color

dark red-purple to red-brown

mp

178-179 °C

solubility

hexane: soluble 100 μg/mL
DMSO: 30 μg/mL
H2O: insoluble

ε (extinction coefficient)

2160-2280 at 478-479 nm in hexane at 1% (vs. 2280, lit.)
2450-2590 at 450-452 nm in hexane at 1% (vs. 2590, lit.)

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C)C(C)(C)CCC1

InChI

1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+

InChI key

OENHQHLEOONYIE-JLTXGRSLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Carotene has been used:
  • as standard for quantification of b-carotene using HPLC analysis
  • as a standard for the identification and quantification of authentic standards for carotenoid groups
  • for standard curve preparation in the measurement of β-carotene production
  • for standard curve preparation in nutritive assays

Biochem/physiol Actions

β-Carotene (pro-vitamin A) is a member of carotenoids. This antioxidant plays a vital role in human physiology. β-Carotene is an abundant carotenoid. It is considered as a precursor of vitamin A, which is a lipid soluble antioxidant with vitamin E properties.
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro. However, in vivo it appears to act either as an antioxidant or a prooxidant depending on cellular environment. It reduces the incidence of many cancers, but enhances lung cancer incidence in smokers.

Caution

Purchase minimum quantity required. Discard one month after opening.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

217.9 °F - closed cup

Flash Point(C)

103.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Liu et al.
Scientific reports, 6, 24117-24117 (2016-04-08)
Protein rational design has become more and more popular for protein engineering with the advantage of biological big-data. In this study, we described a method of rational design that is able to identify desired mutants by analyzing the coevolution of
The nutrient content of five traditional South African dark green leafy vegetables?A preliminary study
Schonfeldt HC and Pretorius B
J. Food Compos. Anal., 24(8), 1141-1146 (2011)
Antioxidant Vitamins and Minerals
Antioxidants in Food, Vitamins and Supplements (2014)
Werner E G Müller et al.
Marine drugs, 10(1), 177-199 (2012-03-01)
The demosponge Suberites domuncula has been described to contain high levels of a proteinaceous toxin, Suberitine, that displays haemolytic activityIn the present study this 7-8 kDa polypeptide has been isolated and was shown to exhibit also cytotoxic effects on cells
Junwei Liu et al.
Journal of experimental botany, 64(7), 1967-1981 (2013-04-10)
Strigolactones (SLs) are newly identified hormones that regulate multiple aspects of plant development, infection by parasitic weeds, and mutualistic symbiosis in the roots. In this study, the role of SLs was studied for the first time in the model plant

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service