Skip to Content
Merck
All Photos(3)

Key Documents

04693132001

Roche

cOmplete, EDTA-free Protease Inhibitor Cocktail

Tablets provided in EASYpacks

Synonym(s):

protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
form:
tablet
solubility:
aqueous buffer: soluble
water: soluble
storage temp.:
2-8°C
Pricing and availability is not currently available.

form

tablet

packaging

pkg of 20 tablets (individually packed in foil blister packs)

manufacturer/tradename

Roche

solubility

aqueous buffer: soluble
water: soluble

absorption

0.08 at 280 nm

storage temp.

2-8°C

General description

cOmplete, EDTA-free Protease Inhibitor Tablets, inhibit a broad spectrum of serine and cysteine proteases. In contrast to other cOmplete tablets they do not contain EDTA - no other chelating agents, e.g. EGTA as well - leaving the stability and the function of metal-dependant proteins unaffected. The affinity purification of Poly-His-tagged fusion proteins using IMAC (Immobilized Metal Affinity Chromatography) is also facilitated (no dialysis necessary).
Due to the optimized composition of the tablets, they show excellent inhibition of serine and cysteine proteases, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete, EDTA-free tablets, contains both irreversible and reversible protease inhibitors. Metallo- and aspartic proteases are not inhibited.

cOmplete EDTA-free Tablets are identical to cOmplete Tablets, the only difference being that no EDTA or other chelating (e.g. EGTA) agents are included.

Specificity

Mixture of several protease inhibitors. For the inhibition of serine, cysteine, but not metalloproteases.

Application

cOmplete, EDTA-free Protease Inhibitor Cocktail has been used as a component in
  • lysis buffer for the lysis of transfected human embryonic kidney (HEK293T) cells and in radioimmunoprecipitation assay (RIPA) lysis buffer for the homogenization of zebrafish embryo[1]
  • tissue protein extraction reagent (T-PER) for the homogenization of hippocampus tissue[2]
  • sodium dodecyl sulphate (SDS) lysis buffer for the lysis of hepatocytes[3]

cOmplete, EDTA-free, efficiently inhibits serine and cysteine proteases in bacterial, yeast, plant, and animal cell extracts.[4][5][6][7] Metallo- and aspartic proteases are not inhibited.
cOmplete, EDTA-free Tablets, are used for the inhibition of proteolytic activity in large volumes (up to 50ml) in which EDTA may interfere with protein stability (e.g., metal-containing proteins) or subsequent assays. Since EDTA interferes with IMAC (Immobilized Metal Affinity Chromatography), cOmplete, EDTA-free is preferentially used in the isolation process of Poly-His-tagged fusion proteins.
If it is necessary to inhibit proteolytic activity in a smaller volume (up to 10ml), we recommend to use cOmplete, Mini, EDTA-free.

One cOmplete, EDTA-free tablet was added per 50 ml incubation solution. Proteolytic activity was determined with the Roche Universal Protease Substrate (casein, resorufin-labeled). When extractions or single-step isolations are necessary in the acid pH range, simply include pepstatin along with cOmplete, EDTA-free tablets to ensure aspartic (acid) protease inhibition. All experiments were performed at +15 to +25°C.

Features and Benefits

Use cOmplete Protease Inhibitor Tablets to protect your proteins from a wide range of proteases. In just minutes, uninhibited proteolytic activity can degrade the protein you have spent days isolating. Each of these convenient water-soluble tablets contains a blend of protease inhibitors that inhibits proteolytic activity from most cell types, including animals, plants, yeast, and bacteria. No weighing or measuring is necessary, ensuring consistent results.
  • Convenience: Simply push the quick-dissolving tablet through the foil packaging of the EASYpack into your buffer.
  • Complete protection: Instantly protect your proteins against a broad range of proteases.
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.
  • Suitable for IMAC purification: Purify poly-histidine tagged proteins with cOmplete His-Tag Purification Resin.

Packaging

20 individually packed cOmplete Protease Inhibitor Cocktail Tablets in foil blisters.

Components

Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Quality

Performance-tested with pancreas extract.

Physical form

Tablets in foil blister packs

Preparation Note

Working concentration: 1 tablet per 50 ml extraction solution
Working solution: Handling

Carefully push the tablet through the foil packaging using the base of your thumb (not fingernail) to prevent the breakage of tablets.

Preparation of Working Solution

One cOmplete EDTA-free tablet is sufficient for the inhibition of the proteolytic activity in 50 ml extraction solution. When very high proteolytic activity is present, one tablet should be used for 25 ml extraction buffer. Tablets can be added directly to the extraction medium. Alternatively, a stock solution (25 X conc.) can be prepared.

Preparation of Stock solution (25 X conc.)

Dissolve one cOmplete EDTA-free tablet in 2 ml dist. water or in 2 ml 100 mM phosphate buffer, pH 7.0.
Storage conditions (working solution): The stock solution can be stored at 2 to 8 °C for 1 to 2 weeks, or at least 12 weeks at -15 to -25 °C.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 25x stock solutions in 2 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1-2 weeks at 2 to 8 °C or at least 12 weeks at -15 to -25 °C; an be used in thiol-containing solutions at 15 to 25 °C.

Storage and Stability

Store at 2 to 8 °C. (storage at -15 to -25 °C is also possible)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Winsel et al.
British journal of cancer, 111(11), 2142-2151 (2014-10-01)
Optimal expression and proper function of key mitotic proteins facilitate control and repair processes that aim to prevent loss or gain of chromosomes, a hallmark of cancer. Altered expression of small regulatory microRNAs is associated with tumourigenesis and metastasis but
Beryl M Jones et al.
eLife, 9 (2020-12-23)
Understanding the regulatory architecture of phenotypic variation is a fundamental goal in biology, but connections between gene regulatory network (GRN) activity and individual differences in behavior are poorly understood. We characterized the molecular basis of behavioral plasticity in queenless honey
Paul D Kasaija et al.
Vaccines, 8(2) (2020-06-24)
Cattle tick infestations and transmitted pathogens affect animal health, production and welfare with an impact on cattle industry in tropical and subtropical countries. Anti-tick vaccines constitute an effective and sustainable alternative to the traditional methods for the control of tick
Density Gradient Centrifugation for Enrichment and Identification of GFP-tagged Chitosomal Microvesicles of Filamentous Fungi.
Bio-protocol null
Yangtian Yan et al.
Molecular medicine reports, 22(5), 3886-3894 (2020-10-02)
Emerging evidence has indicated that long non‑coding RNAs (lncRNAs) are closely associated with the pathogenesis of ischemic stroke. It has been reported that small nucleolar RNA host gene 12 (SNHG12) serves a critical role in ischemic stroke by acting as a

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service