Skip to Content
Merck
All Photos(1)

Key Documents

11666975001

Roche

HA Peptide

9-amino acid synthetic peptide from hemagglutinin (HA) of an influenza virus

Synonym(s):

peptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
Pricing and availability is not currently available.

Assay

95% (HPLC)

form

lyophilized

packaging

pkg of 5 mg

manufacturer/tradename

Roche

storage temp.

−20°C

Related Categories

General description

HA (hemagglutinin) is a surface glycoprotein, expressed by influenza virus.[1]

Application

HA Peptide has been used in the following :
  • Protein purification from Baculovirus infected cells.[2]
  • Affinity chromatography.[3][4]
  • Chromatin immunoprecipitation.[5][6]

Biochem/physiol Actions

HA (hemagglutinin) protein is responsible for the infectivity of human influenza by inducing the production of neutralizing antibodies. HA mediates viral attachment to the surface of oligosaccharide receptors (at sialic acid terminal).[7] HA contains conserved cross-reactive antigenic determinants, which is predicted to stimulate cross-protective antibodies.[1]

Quality

  • Purity: =95% (HPLC)
  • Function test: Competitive ELISA, using Anti-HA-coated microplate to measure free HA Peptide inhibition of HA-peroxidase binding with detection by ABTS.

Reconstitution

Add 5 ml TBS to get a final concentration of 1 mg/ml.
Rehydrate for at least 10 minutes.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A common neutralizing epitope conserved between the hemagglutinins of influenza A virus H1 and H2 strains.
Okuno Y, et al.
Journal of Virology, 67(5), 2552-2558 (1993)
Physical and genetic associations of the Irc20 ubiquitin ligase with Cdc48 and SUMO.
Richardson A, et al.
PLoS ONE, 8(10), e76424-e76424 (2013)
R J Riese et al.
Immunity, 4(4), 357-366 (1996-04-01)
Destruction of li by proteolysis is required for MHC class II molecules to bind antigenic peptides, and for transport of the resulting complexes to the cell surface. The cysteine protease cathepsin S is highly expressed in spleen, lymphocytes, monocytes, and
Spt10 and Swi4 control the timing of histone H2A/H2B gene activation in budding yeast.
Eriksson P R, et al.
Molecular and Cellular Biology, 31(3), 557-572 (2011)
Creation and characterization of BAC-transgenic mice with physiological overexpression of epitope-tagged RCAN1 (DSCR1).
Xing L, et al.
Mammalian Genome, 24(1-2), 30-43 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service