Skip to Content
Merck

E6779

EZview Red Anti-HA Affinity Gel

Synonym(s):

Anti-HA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
12352203
Technique(s):
affinity chromatography: suitable, immunoprecipitation (IP): suitable
Species reactivity:
-
Application:
IP, affinity chromatography
Citations:
242
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

1 yr at 2‑8 °C

Quality Level

analyte chemical class(es)

proteins (hemagglutinin)

technique(s)

affinity chromatography: suitable, immunoprecipitation (IP): suitable

matrix

crosslinked agarose

capacity

≥0.4 mg/mL, gel binding capacity (HA-tagged fusion protein)

storage temp.

2-8°C

General description

EZview Red Anti-HA Affinity Gel is a red colored Anti-HA agarose affinity gel that contains anti-HA monoclonal antibody which is covalently attached to crosslinked agarose beads. The HA tag has the sequence YPYDVPDYA and derives from the hemagglutinin (HA) protein. This sequence occurs at amino acid residues 98-106 of the full-length HA protein. The HA tag has a high presence of charged amino acids, which enhances its likelihood of strong antibody recognition.

Application

EZview Red Anti-HA Affinity Gel has been used for immunoblot analysis and immunoprecipitation assay.
Suitable for Affinity capture (immunoprecipitation) of N- or C- terminal HA-tagged fusion proteins from E. coli or mammalian cell.
When performing small-scale affinity capture, such as immunoprecipitation, the affinity matrix is difficult to see in the microcentrifuge tubes. Accidental aspiration of the resin leads to quantitative variability in results. The EZview Red Affinity Gels greatly reduces the risk of pellet loss. EZview resins perform as well as conventional non-colored affinity gels, but allow the user to easily differentiate pellet from supernatant. This correlates to more accurate data because less protein is lost.

Biochem/physiol Actions

Binding specificity: HA-tag peptide (N-Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala-C). It recognizes reduced forms of HA-tagged proteins and is reactive with N or C-terminal HA-tagged fusion proteins.

Features and Benefits

  • Increased visibility - Red color reduces risk of incidental aspiration
  • Improved recovery of target protein by reduced accidental loss
  • Higher reproducibility - More consistent yields

Physical form

1:1 (v/v) suspension in PBS containing 50% glycerol and 15 ppm Kathon

Other Notes

The affinity gel contains affinity purified anti-HA mouse monoclonal antibody (Clone HA-7) covalently attached to beaded agarose. The HA tag corresponds to amino acid residues 98-106 (YPYDVPDYA) of human influenza virus hemagglutinin (HA).

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


The nonreceptor tyrosine kinase c-Abl phosphorylates Runx1 and regulates Runx1-mediated megakaryocyte maturation
Liu H, et al.
Biochimica et biophysica acta. General subjects, 1865(8), 1060-1072 (2018)
Activation of c-Abl kinase potentiates the anti-myeloma drug lenalidomide by promoting DDA1 protein recruitment to the CRL4 ubiquitin ligase
Gao S, et al.
The Journal of biological chemistry, 292(9), 3683-3691 (2017)
Novel Mitochondrial Substrates of Omi Indicate a New Regulatory Role in Neurodegenerative Disorders
Johnson,F et al.
PLoS Biology, 4(9) (2009)



Global Trade Item Number

SKUGTIN
E6779-5X1ML04061833604656
E6779-1ML04061838099273
E6779-500UL04061838248541