Skip to Content
Merck
All Photos(1)

Key Documents

8.40130

Sigma-Aldrich

4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl

(free radical) for synthesis

Synonym(s):

4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Hydroxy-TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18NO2
CAS Number:
Molecular Weight:
172.24
MDL number:
UNSPSC Code:
12352119
EC Index Number:
218-760-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

crystals

autoignition temp.

260 °C (DIN 51794)

potency

1053 mg/kg LD50, oral (Rat)

pH

8.2 (20 °C, 20 g/L in H2O)

mp

67-71 °C

transition temp

flash point 146 °C (DIN 51758)

solubility

1670 g/L

storage temp.

2-30°C

SMILES string

[N+]1(C(CC(CC1(C)C)O)(C)C)[O-]

InChI

1S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11H,5-6H2,1-4H3

InChI key

UZFMOKQJFYMBGY-UHFFFAOYSA-N

Application


  • Vitamin C Determination: A study that utilizes TEMPO for a simple and rapid determination of vitamin C content in various fruits, showcasing an economical approach to antioxidant testing (Ihara et al., 2017).

  • Herbicide Development: A study on the synthesis of phenoxy carboxylic herbicides based on the piperidine scaffold, where TEMPO derivatives showed unexpected fungicidal activity (Zakrzewski & Krawczyk, 2014).


Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 67 °C
Melting range (upper value): ≤ 71 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

294.8 °F - DIN 51758

Flash Point(C)

146 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service