Skip to Content
Merck
All Photos(2)

Key Documents

115746

Sigma-Aldrich

2,2,6,6-Tetramethyl-4-piperidinol

98%

Synonym(s):

4-Hydroxy-2,2,6,6-tetramethylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO
CAS Number:
Molecular Weight:
157.25
Beilstein:
105039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

bp

212-215 °C (lit.)

mp

129-131 °C (lit.)

functional group

hydroxyl

SMILES string

CC1(C)CC(O)CC(C)(C)N1

InChI

1S/C9H19NO/c1-8(2)5-7(11)6-9(3,4)10-8/h7,10-11H,5-6H2,1-4H3

InChI key

VDVUCLWJZJHFAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,6,6-Tetramethyl-4-piperidinol has been used to study the green and efficient method for chemoselective deoxidization of graphene oxide via the ultraviolet irradiation[1].
2,2,6,6-tetramethyl-4-piperidinol has been used to study the irradiation of Camptothecin (CPT) in aerated dimethylsulfoxide (DMSO) solution that leads to formation of singlet oxygen[2].
Used as:
  • An additive for polymer modified hindered amine light stabilizers
  • A light stabilizer for polyamide 66 fibers containing acid blue dyes

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hirohisa Sato et al.
The Journal of toxicological sciences, 41(6), 793-799 (2016-11-18)
The present study aimed to evaluate the acute locally injurious property of our most current hydroxyl radical generation system by hydrogen peroxide (H2O2) photolysis. This system, which releases 3% H2O2 with a 405-nm laser, was developed in our laboratory for
Keisuke Nakamura et al.
PloS one, 8(3), e60053-e60053 (2013-03-26)
In the present study, the bactericidal effect of photo-irradiated proanthocyanidin was evaluated in relation to reactive oxygen species formation. Staphylococcus aureus suspended in proanthocyanidin aqueous solution was irradiated with light from a laser at 405 nm. The bactericidal effect of
Fei Zhao et al.
ACS nano, 6(4), 3027-3033 (2012-03-17)
We report a green and efficient method for chemoselective deoxidization of graphene oxide via the ultraviolet irradiation catalyzed with 2,2,6,6-tetramethyl-4-piperidinol. While the sp(2)-hybridized oxygen functional groups are removed after the reduction, the epoxy and hydroxyl groups are retained in the
Shishu Zhu et al.
Environmental science & technology, 53(1), 307-315 (2018-11-28)
Minerals and transitional metal oxides of earth-abundant elements are desirable catalysts for in situ chemical oxidation in environmental remediation. However, catalytic activation of peroxydisulfate (PDS) by manganese oxides was barely investigated. In this study, one-dimension manganese dioxides (α- and β-MnO2)
Kouichi Nakagawa et al.
Journal of oleo science, 66(3), 315-319 (2017-02-14)
This study investigated the location and distribution of paramagnetic species in apple seeds using electron paramagnetic resonance (EPR) and X-band (9 GHz) EPR imaging (EPRI). EPR primarily detected two paramagnetic species per measured seed. These two different radical species were

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service