Skip to Content
Merck
All Photos(3)

Key Documents

163945

Sigma-Aldrich

4-Amino-TEMPO, free radical

97%

Synonym(s):

4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidinyloxy, free radical, 4-Amino-TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19N2O
CAS Number:
Molecular Weight:
171.26
Beilstein:
3933966
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

~0 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)CC(N)CC(C)(C)N1[O]

InChI

1S/C9H19N2O/c1-8(2)5-7(10)6-9(3,4)11(8)12/h7H,5-6,10H2,1-4H3

InChI key

XUXUHDYTLNCYQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful spin label for studying biological systems, polymers, and as a building block for more elaborate spin labels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Naber et al.
Biochemistry, 33(13), 3855-3861 (1994-04-05)
Each actin molecule contains a nucleotide, tightly bound in a deep cleft that divides the molecule. To probe conformational changes within this region of the molecule, we have incorporated two spin label analogues of ATP into actin. In both analogs
Spin-Labeled Dendrimers in EPR Imaging with Low Molecular Weight Nitroxides.
Alexander T. Yordanov et al.
Angewandte Chemie (International ed. in English), 40(14), 2690-2692 (2001-07-18)
Darja Jaušovec et al.
Carbohydrate polymers, 116, 74-85 (2014-12-03)
The chemo-enzymatic modification of cellulose nanofibers (CNFs) using laccase as biocatalysts and TEMPO or 4-Amino-TEMPO as mediators under mild aqueous conditions (pH 5, 30 °C) has been investigated to introduce surface active aldehyde groups. 4-Amino TEMPO turned out to be
Boris V Trubitsin et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 163(2), 257-269 (2003-08-14)
We present a method for measuring the transmembrane pH difference (deltapH=pHin-pHout) in chloroplasts with a spin label TEMPAMINE (4-amino-2,2,6,6-tetramethylpiperidine-1-oxyl) accumulating inside the thylakoids in response to generation of deltapH. Experiments with chloroplasts suspended in the media of different osmolarity demonstrated
Jianfei Jiang et al.
The Journal of pharmacology and experimental therapeutics, 320(3), 1050-1060 (2006-12-21)
Suppression of mitochondrial production of reactive oxygen species is a promising strategy against intrinsic apoptosis typical of degenerative diseases. Stable nitroxide radicals such as 4-hydroxy-2,2,6,6-tetramethyl piperidine-1-oxyl (TEMPOL) and its analogs combine several important features, including recycleability, electron acceptance from respiratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service