Skip to Content
Merck
All Photos(5)

Documents

214000

Sigma-Aldrich

TEMPO

greener alternative

98%

Synonym(s):

2,2,6,6-Tetramethylpiperidine 1-oxyl, 2,2,6,6-Tetramethyl-1-piperidinyloxy, free radical, TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18NO
CAS Number:
Molecular Weight:
156.25
Beilstein:
1422418
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

36-38 °C (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

CC1(C)CCCC(C)(C)N1[O]

InChI

1S/C9H18NO/c1-8(2)6-5-7-9(3,4)10(8)11/h5-7H2,1-4H3

InChI key

QYTDEUPAUMOIOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TEMPO (2,2,6,6-Tetramethylpiperidin-1-oxyl) is a stable free radical commonly used as an oxidizing agent in organic synthesis. Copper/TEMPO has been utilized in the catalytic oxidation of primary alcohols to aldehydes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

TEMPO can be used:
  • As a catalytic oxidant for copper-catalyzed, greener oxidation of alcohols under aerobic conditions.
  • As a catalytic oxidant in the iodobenzene diacetae oxidation of nerol to neral.
  • For trapping the styrenyl radical generated from benzoyl peroxide during nitroxide-mediated radical polymerization of styrene.
  • In the carboxylation of water-resistant nanofibrillated cellulose (NFC) films.

Copper(I)/TEMPO-catalyzed aerobic oxidation of primary alcohols to aldehydes with ambient air

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geisslmeir, David; et al.
Monatshefte fur Chemie / Chemical Monthly, 136(9), 1591-1599 (2005)
Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator.
Lee NS and Wooley KL.
Material Matters, 5, 8-15 (2010)
Wan Ru Leow et al.
Science (New York, N.Y.), 368(6496), 1228-1233 (2020-06-13)
Chemicals manufacturing consumes large amounts of energy and is responsible for a substantial portion of global carbon emissions. Electrochemical systems that produce the desired compounds by using renewable electricity offer a route to lower carbon emissions in the chemicals sector.
Soham Maity et al.
Journal of the American Chemical Society, 135(9), 3355-3358 (2013-02-15)
Nitroolefin is a common and versatile reagent. Its synthesis from olefin is generally limited by the formation of mixture of cis and trans compounds. Here we report that silver nitrite (AgNO2) along with TEMPO can promote the regio- and stereoselective
Organic Syntheses, 83, 18-18 (2006)

Related Content

he Stahl Lab focuses on the development of catalysts and catalytic reactions for selective oxidation of organic molecules, with particular emphasis on aerobic oxidation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service