Skip to Content
Merck
All Photos(1)

Key Documents

8.02954

Sigma-Aldrich

N,N′-Dicyclohexylcarbodiimide

for peptide synthesis

Synonym(s):

N,N′-Dicyclohexylcarbodiimide, DCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(NC6H11)2
CAS Number:
Molecular Weight:
206.33
MDL number:
UNSPSC Code:
12352111
EC Index Number:
208-704-1
NACRES:
NA.22

Product Name

N,N′-Dicyclohexylcarbodiimide, for synthesis

Quality Level

form

solid

potency

1110 mg/kg LD50, oral (Rat)
71 mg/kg LD50, skin (Rat)

reaction suitability

reaction type: Coupling Reactions

bp

148-152 °C/15 hPa

mp

35-36 °C

transition temp

flash point 113 °C

density

0.95 g/cm3 at 40 °C

bulk density

920 kg/m3

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N(=C=NC2CCCCC2)C1CCCCC1

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

General description

N,N′-Dicyclohexylcarbodiimide (DCC) is a highly versatile dehydrating reagent with varied applications in organic chemistry. It is most frequently used as a coupling agent for the synthesis of amides, esters and thioesters from carboxylic acids and amines, alcohol and thiols, respectively. In peptide synthesis applications, DCC is used in conjunction with auxiliary nucleophiles such as Oxyma Pure and HOBt, to minimize side reactions such as racemization, N-acylurea or oxazolone formation. It is used as a dehydration reagent in the Moffatt oxidation of primary alcohols to aldehydes and in the conversion of amides to nitriles. The by-product of the dehydration reaction, dicyclohexylurea, is sparingly soluble in most reaction solvents and can be removed by filtration.

Application

Recent applications of N,N′-Dicyclohexylcarbodiimide include:
  • The synthesis of optically pure N-acyl-N,N′-dicyclohexylureas.
  • The activation of the carboxylic acid groups in aromatic carboxylic acids to facilitates their reaction with (N-isocyanimino)trifluoroacetamide to form the corresponding 1,3,4-oxadiazole derivatives.
  • The synthesis of poly (vinyl alcohol-co-vinyl levulinate) copolymers for use in biomedical applications.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N, N-dicyclohexylcarbodiimide assisted synthesis and characterization of poly (vinyl alcohol-co-vinyl levulinate
Wang YM, et al.
Polymer, 9793-9802 (2005)
Synthesis of sterically congested 1, 3, 4-oxadiazole derivatives from aromatic carboxylic acids, N, N-dicyclohexylcarbodiimide and (N-isocyanimino) triphenylphosphorane
Nasrabadi FZ, et al.
Molecular Diversity, 791-798 (2011)
Dicyclohexylcarbodiimide
Albert, J. S. et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-9 (2017)
Formation of optically pure N-acyl-N, N?-dicyclohexylurea in N, N?-dicyclohexylcarbodiimide-mediated peptide synthesis
SLEBIODA, M, et al.
International Journal of Peptide and Protein Research, 35, 539-541 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service