Skip to Content
Merck
All Photos(5)

Key Documents

130672

Sigma-Aldrich

N-Hydroxysuccinimide

98%

Synonym(s):

1-Hydroxy-2,5-pyrrolidinedione, HOSu, NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO3
CAS Number:
Molecular Weight:
115.09
Beilstein:
113913
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

95-98 °C (lit.)

functional group

imide

SMILES string

ON1C(=O)CCC1=O

InChI

1S/C4H5NO3/c6-3-1-2-4(7)5(3)8/h8H,1-2H2

InChI key

NQTADLQHYWFPDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Hydroxysuccinimide can be used in the synthesis of N-hydroxysuccinimide ester via dehydration reaction in the presence of dicyclohexylcarbodiimide in carboxylic acid.
N-Hydroxysuccinimide has been used in the synthesis of intermediates such as:
  • N

  • -succinimidyl 3-(di-tert-butylfluorosilyl)benzoate
  • 4-[2,2-bis[(p-tolylsulfonyl)-methyl] acetyl]benzoic acid-NHS ester
  • N-succinimidyl 3-iodobenzoate

It has also been used in a protocol for the surface modification of microchannels of a microfluidic-integrated surface plasmon resonance (SPR) platform for the detection and quantification of bacterial pathogens.
Additive used in the carbodiimide method for improved amidations and peptide couplings.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PEGylation of native disulfide bonds in proteins.
Brocchini S
Nature Protocols, 1(5), 2241-2241 (2006)
Zbigniew Czajgucki et al.
Journal of peptide science : an official publication of the European Peptide Society, 12(10), 653-662 (2006-07-19)
Edeines are pentapeptide amide antibiotics composed of four nonprotein amino acids, glycine, and polyamine. They exhibit antimicrobial and immunosuppressive activities and are universal inhibitors of translation. Moreover, it was proven that the free ionizable carboxy group in the (2R, 6S
Pico Y et al.
Food Toxicants Analysis: Techniques, Strategies and Developments (2007)
Cong Li et al.
Journal of the American Chemical Society, 128(47), 15072-15073 (2006-11-23)
The conjugate of bacterial cytosine deaminase (bCD) and poly-l-lysine (PLL) that was functionalized with biotin, rhodamine, and Gd3+-DOTA was synthesized and characterized. It demonstrated high relaxivity, improved enzymatic specificity to prodrug 5-fluorocytosine, low cytotoxicity, efficient cell uptake, and high enzymatic
D Sehgal et al.
Analytical biochemistry, 218(1), 87-91 (1994-04-01)
Water-soluble carbodiimides are widely used for carboxyl-amine conjugation. However, extremely variable and low yields, obtained under a variety of conditions, have been a serious problem in the coupling. A simple method, optimizing various parameters of the coupling reaction, in which

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service